Merck
  • Synthesis of bis-acyclonucleoside analogues bearing benzothienyl-1,2,4-Triazol-3-Yl-disulfide under conventional and microwave methods.
Synthesis of bis-acyclonucleoside analogues bearing benzothienyl-1,2,4-Triazol-3-Yl-disulfide under conventional and microwave methods.
Nucleosides, nucleotides & nucleic acids (2013-01-31)
Mohamed R Aouad, Nadjet Rezki, Mouslim Messali, El Sayed H El Ashry
ZUSAMMENFASSUNG
The oxidation of 5-(3-chlorobenzo[b]thien-2-yl)-4H-1,2,4-triazole-3-thiol (1) with a solution of iodine and potassium iodide at room temperature afforded [5-(3-chlorobenzo[b]thien-2-yl)-4H-1,2,4-triazole-3-yl]disulfide (2). In contrast, when the reaction mixture was heated or irradiated by MW, an unexpected additional product was obtained and identified as 3-(3-chlorobenzo[b]thien-2-yl)-4H-1,2,4-triazole (3); the ratio of products was 3:1. The preferred conformer of 2 was deduced from the theoretical calculation. The alkylation of compounds 2 and 3 with epichlorohydrin and hydroxyalkylating agents gave the corresponding N,N-bis- and N-acyclonucleosides analogues 8-15.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N,N-Dimethylformamid, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamid, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Kaliumiodid, ACS reagent, ≥99.0%
Sigma-Aldrich
N,N-Dimethylformamid, anhydrous, 99.8%
Sigma-Aldrich
Kaliumcarbonat, ACS reagent, ≥99.0%
Sigma-Aldrich
Kaliumiodid, ReagentPlus®, 99%
Sigma-Aldrich
N,N-Dimethylformamid, ReagentPlus®, ≥99%
Sigma-Aldrich
Kaliumcarbonat, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
N,N-Dimethylformamid, Molecular Biology, ≥99%
Sigma-Aldrich
Kaliumcarbonat, reagent grade, ≥98%, powder, −325 mesh
Sigma-Aldrich
N,N-Dimethylformamid, biotech. grade, ≥99.9%
Sigma-Aldrich
Kaliumcarbonat, ReagentPlus®, 99%
Sigma-Aldrich
Kaliumiodid, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Goldätzmittel, Standard
Sigma-Aldrich
Kaliumcarbonat, 99.995% trace metals basis
Sigma-Aldrich
Kaliumcarbonat, BioXtra, ≥99.0%
Sigma-Aldrich
Kaliumcarbonat, powder, 99.99% trace metals basis
Sigma-Aldrich
Kaliumiodid, ≥99.99% trace metals basis
Supelco
N,N-Dimethylformamid, analytical standard
Sigma-Aldrich
Kaliumiodid, suitable for plant cell culture
Sigma-Aldrich
Kaliumiodid, puriss., meets analytical specification of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Kaliumiodid, BioXtra, ≥99.0%
Sigma-Aldrich
Kaliumcarbonat, meets USP testing specifications
Sigma-Aldrich
Kaliumcarbonat, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Kaliumiodid, AnhydroBeads, −10 mesh, 99.998% trace metals basis