Merck
  • DNA binding, DNA cleavage, antioxidant and cytotoxicity studies on ruthenium(II) complexes of benzaldehyde 4-methyl-3-thiosemicarbazones.
DNA binding, DNA cleavage, antioxidant and cytotoxicity studies on ruthenium(II) complexes of benzaldehyde 4-methyl-3-thiosemicarbazones.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2013-02-05)
Krishnan Sampath, Subbaiyan Sathiyaraj, Chinnasamy Jayabalakrishnan
ZUSAMMENFASSUNG
Four new ruthenium(II) complexes with N(4)-methyl thiosemicarbazone ligands, (E)-2-(2-chlorobenzylidene)-N-methylhydrazinecarbothioamide (HL(1)) and (E)-N-methyl-2-(2-nitrobenzylidene)hydrazinecarbothioamide (HL(2)), were prepared and fully characterized by various spectro-analytical techniques. The Schiff bases act as bidentate, monobasic chelating ligands with S and N as the donor sites and are preferably found in the thiol form in all the complexes studied. The molecular structure of HL(1) and HL(2) were determined by single crystal X-ray diffraction method. DNA binding of the compounds was investigated by absorption spectroscopy which indicated that the complexes bind to DNA via intercalation. The oxidative cleavage of the complexes with CT-DNA inferred that the effects of cleavage are dose dependent. Antioxidant studies of the ligands and complexes showed the significant antioxidant activity against DPPH radical. In addition, the in vitro cytotoxicity of the ligands and complexes against MCF-7 cell line was assayed which showed higher cytotoxic activity with the lower IC(50) values indicating their efficiency in killing the cancer cells even at low concentrations.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Benzaldehyd, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzaldehyd, purified by redistillation, ≥99.5%
Supelco
Benzaldehyd, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Desoxyribonukleinsäure aus Kalbsthymus, Type XV, Activated, lyophilized powder
Sigma-Aldrich
Benzaldehyd, ≥98%, FG, FCC
Sigma-Aldrich
Benzaldehyd, puriss. p.a., ≥99.0% (GC)
Supelco
Benzaldehyd, analytical standard
Sigma-Aldrich
Benzaldehyd, natural, FCC, FG
Sigma-Aldrich
Desoxyribonukleinsäure, einsträngig aus Kalbsthymus, lyophilized powder
Sigma-Aldrich
Desoxyribonukleinsäure aus Kalbsthymus, Genomic, unsheared
Benzaldehyd, European Pharmacopoeia (EP) Reference Standard