Merck
  • Effect of curing light distance and different mouthwashes on the sorption and solubility of a nanofilled composite.
Effect of curing light distance and different mouthwashes on the sorption and solubility of a nanofilled composite.
The European journal of esthetic dentistry : official journal of the European Academy of Esthetic Dentistry (2013-02-08)
Diogo de Azevedo Miranda, Carlos Eduardo dos Santos Bertoldo, Glaucia Maria Bovi Ambrosano, Flávio Henrique Baggio Aguiar, Débora Alves Nunes Leite Lima, José Roberto Lovadino
ZUSAMMENFASSUNG
The aim of this in vitro study was to evaluate the sorption (Sp) and solubility (Sl) of a nanofilled composite (Filtek™ Z350) light-activated from the top in three curing light distances (0 mm, 3 mm and 6 mm) and immersed in three different mouthwashes (Plax Fresh Mint, Plax Alcohol Free, and Plax Whitening). Specimens (6 mm Ø and 1 mm height) were prepared (n = 5) and initially the degree of conversion (DC%) of discs was obtained by using a fourier transform infrared spectrometer. Then, the specimens were placed in a desiccator at 37°C and were weighed until a constant mass (m1) was obtained. The discs were immersed in the mouthwashes for 30 days. Twice a day, the samples were immersed in 2 ml of the mouthwashes (12 hr intervals). After the 30-day period, the discs were reweighed (m2). The reconditioning in the desiccator was done until a constant mass (m3) was obtained. The data of DC% were analyzed by analysis of variance (ANOVA) in split plots and the results showed statistical difference when photoactivated to 6 mm from the tip of the curing unit. The sorption data were submitted to two-way ANOVA and Tukey's Test at a 5% level of significance and it was observed that a statistical difference (P < 0.05) was only cured when the distance was 6 mm, but there was no difference in sorption between mouthwashes. The solubility data were analyzed by Kruskal-Wallis's Test (P = 0.05) and Dunn's Test, since a non-normal data distribution was observed. The values were negative, which means that there was a mass gain, masking the real solubility.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Supelco
Natriumdodecylsulfat, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Triclosan, 97.0-103.0% (active substance, GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Natriumdodecylsulfat, ≥90% ((Assay))
Sigma-Aldrich
Natriumdodecylsulfat, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri