Merck
Directly probing the effects of ions on hydration forces at interfaces.
Journal of the American Chemical Society (2013-02-13)
Jason I Kilpatrick, Siu-Hong Loh, Suzanne P Jarvis
ZUSAMMENFASSUNG
Understanding the influence of water layers adjacent to interfaces is fundamental in order to fully comprehend the interactions of both biological and nonbiological materials in aqueous environments. In this study, we have investigated hydration forces at the mica-electrolyte interface as a function of ion valency and concentration using subnanometer oscillation amplitude frequency modulation atomic force microscopy (FM-AFM). Our results reveal new insights into the nature of hydration forces at interfaces due to our ability to measure high force gradients without instability and in the absence of lateral confinement due to the use of an atomically sharp tip. We demonstrate the influence of electrolytes on the properties of both primary and structural hydration forces and reveal new insights into the interplay between these phenomena in determining the interaction forces experienced by a nanoscale object approaching an interface. We also highlight the difficulty in directly comparing hydration force data from different measurement techniques where the nature of the perturbation induced by differing interaction geometries is likely to dramatically affect the results.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
Magnesiumchlorid, powder, <200 μm
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesiumchlorid, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesiumchlorid -Lösung, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesiumchlorid -Lösung, 0.1 M
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~0.025 M in H2O