Merck
  • Calcium alginate beads encapsulated PMMA-g-CS nano-particles for α-chymotrypsin immobilization.
Calcium alginate beads encapsulated PMMA-g-CS nano-particles for α-chymotrypsin immobilization.
Carbohydrate polymers (2013-02-13)
M A Abd El-Ghaffar, M S Hashem
ZUSAMMENFASSUNG
Chitosan grafted with polymethyl methacrylate (PMMA-g-CS) was prepared via a free-radicals polymerization technique as a carrier for enzyme immobilization. α-Chymotrypsin (CT), as an enzyme model in this study, was immobilized onto the prepared PMMA-g-CS via covalent bonding. Calcium alginate (CA) beads were developed for encapsulating PMMA-g-CS-CT to produce PMMA-g-CS-CT/CA composite beads. Morphology and size of PMMA-g-CS particles were investigated by TEM and found to be in the nanoscale. The structure and surface morphology of the beads before and after immobilization process were characterized by FT-IR and SEM, respectively. Both the bound CT content and relative activity of immobilized enzyme were measured. A higher retained activity (about 97.7%) obtained for the immobilized CT at pH 9 for 24 h. The results indicated that immobilized CT maintained excellent performance even after 25 reuses and retained 75% from its original activity after 60 days of storage at 25 °C.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumalginat, powder
Sigma-Aldrich
Natriumalginat
Sigma-Aldrich
αα-Chymotrypsin aus Rinderpankreas, Type II, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
Natriumalginat aus Braunalgen, Medium viscosity
Sigma-Aldrich
Natriumalginat aus Braunalgen, low viscosity
Sigma-Aldrich
Natriumalginat aus Braunalgen, BioReagent, suitable for immobilization of micro-organisms
Sigma-Aldrich
Alginsäure aus Braunalgen, powder
Sigma-Aldrich
αα-Chymotrypsin, (TLCK treated to inactivate residual tryspin activity), Type VII, essentially salt-free, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
αα-Chymotrypsin aus Rinderpankreas, Type I-S, essentially salt-free, lyophilized powder
Sigma-Aldrich
αα-Chymotrypsin aus Rinderpankreas, ≥40 units/mg protein, vial of 5 mg
Sigma-Aldrich
Calciumalginat aus Braunalgen
Sigma-Aldrich
αα-Chymotrypsin aus Rinderpankreas, suitable for protein sequencing, salt-free, lyophilized powder
Sigma-Aldrich
α-Chymotrypsin from human pancreas, lyophilized powder