Merck
  • Kinetic and anion inhibition studies of a β-carbonic anhydrase (FbiCA 1) from the C4 plant Flaveria bidentis.
Kinetic and anion inhibition studies of a β-carbonic anhydrase (FbiCA 1) from the C4 plant Flaveria bidentis.
Bioorganic & medicinal chemistry letters (2013-02-19)
Simona Maria Monti, Giuseppina De Simone, Nina A Dathan, Martha Ludwig, Daniela Vullo, Andrea Scozzafava, Clemente Capasso, Claudiu T Supuran
ZUSAMMENFASSUNG
Several β-carbonic anhydrases (CAs, EC 4.2.1.1) are present in all land plants examined thus far. Here we report the first detailed biochemical characterization of one such isoform, FbiCA 1, from the C4 plant Flaveria bidentis, which was cloned, purified and characterized as recombinant protein. FbiCA 1 has an interesting CO2 hydrase catalytic activity (kcat of 1.2×10(5) and kcat/Km of 7.5×10(6)M(-1)×s(-1)) and was moderately inhibited by most simple/complex inorganic anions. Potent FbiCA 1 inhibitors were also detected, such as trithiocarbonate, diethyldithiocarbamate, sulfamide, sulfamic acid, phenylboronic acid and phenylarsonic acid (KIs in the range of 4-60μM). Such inhibitors may be used as tools to better understand the role of various β-CA isoforms in photosynthesis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Carboanhydrase aus Rindererythrocyten, lyophilized powder, ≥2,000 W-A units/mg protein
Sigma-Aldrich
Carboanhydrase aus Rindererythrocyten, ≥95% (SDS-PAGE), specific activity ≥3,500 W-A units/mg protein, lyophilized powder
Sigma-Aldrich
Carboanhydrase aus Rindererythrocyten, BioReagent, suitable for GFC marker
Sigma-Aldrich
Karbonanhydraseisozym II aus Rindererythrocyten, lyophilized powder, ≥3,000 W-A units/mg protein
Sigma-Aldrich
Carboanhydrase aus Rindererythrocyten, For use as a marker in SDS-PAGE
Sigma-Aldrich
Kohlensäureanhydrase I aus Humanerythrocyten
Sigma-Aldrich
Carboanhydrase aus Rindererythrocyten, non-denaturing PAGE marker
Sigma-Aldrich
Carbonic Anhydrase Isozyme II human, ≥80%, powder, ≥3,000 W-A units/mg protein
Sigma-Aldrich
Karbonanhydraseisozym II aus Rindererythrocyten, lyophilized powder, ≥2,000 W-A units/mg protein
Supelco
Kohlensäureanhydrase I aus Humanerythrocyten, Isoelectric focusing marker, pI 6.6
Sigma-Aldrich
Karbonanhydraseisozym II aus Rindererythrocyten, Isoelectric focusing marker, pI 5.4