Merck
  • Fragment-based discovery of 8-hydroxyquinoline inhibitors of the HIV-1 integrase-lens epithelium-derived growth factor/p75 (IN-LEDGF/p75) interaction.
Fragment-based discovery of 8-hydroxyquinoline inhibitors of the HIV-1 integrase-lens epithelium-derived growth factor/p75 (IN-LEDGF/p75) interaction.
Journal of medicinal chemistry (2013-03-01)
Erik Serrao, Bikash Debnath, Hiroyuki Otake, Yuting Kuang, Frauke Christ, Zeger Debyser, Nouri Neamati
ZUSAMMENFASSUNG
On the basis of an initial molecular modeling study suggesting the favorable binding of the "privileged" fragment 8-hydroxyquinoline with HIV-1 integrase (IN) at the IN-lens epithelium-derived growth factor/p75 (LEDGF/p75) interface , we developed a set of modified 8-hydroxyquinoline fragments demonstrating micromolar IC50 values for inhibition of the IN-LEDGF/p75 interaction, but significant cytotoxicity was associated with these initial compounds. Diverse modifications at the C5 and C7 carbons of the 8-hydroxyquinoline core improved potency, but reduction of diversity to only modifications at the C5 position ultimately yielded potent inhibitors with low cytotoxicity. Two of these particular compounds, 5-((p-tolylamino)methyl)quinolin-8-ol and 5-(((3,4-dimethylphenyl)amino)methyl)quinolin-8-ol, inhibited viral replication in MT-4 cells with low micromolar EC50. This is the first study providing evidence for 8-hydroxyquinolines as novel inhibitors of the IN-LEDGF/p75 interaction. Our lead compounds are druglike, have low molecular weights, and are amenable to various substitutions suitable for enhancing their potency and selectivity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Piperidin, ReagentPlus®, 99%
Sigma-Aldrich
Piperidin, ≥99.5%, purified by redistillation
Sigma-Aldrich
Piperazin, ReagentPlus®, 99%
Sigma-Aldrich
8-Hydroxychinolin, ACS reagent, 98.5%
Sigma-Aldrich
Piperidin, biotech. grade, ≥99.5%
Sigma-Aldrich
Piperazin, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
8-Hydroxychinolin
Supelco
Piperazin Hexahydrat, analytical standard
Sigma-Aldrich
Piperidin -Lösung, suitable for peptide synthesis, 20% in DMF
Sigma-Aldrich
8-Hydroxychinolin, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (perchloric acid titration)
Supelco
8-Hydroxychinolin, PESTANAL®, analytical standard