- Carbonic anhydrases.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Carboanhydrase aus Rindererythrocyten, lyophilized powder, ≥2,000 W-A units/mg protein
Sigma-Aldrich
Carboanhydrase aus Rindererythrocyten, ≥95% (SDS-PAGE), specific activity ≥3,500 W-A units/mg protein, lyophilized powder
Sigma-Aldrich
Karbonanhydraseisozym II aus Rindererythrocyten, lyophilized powder, ≥3,000 W-A units/mg protein
Sigma-Aldrich
Karbonanhydraseisozym II aus Rindererythrocyten, lyophilized powder, ≥2,000 W-A units/mg protein
Sigma-Aldrich
Karbonanhydraseisozym II aus Rindererythrocyten, Isoelectric focusing marker, pI 5.4