Merck
  • Vasodilator activity of the essential oil from aerial parts of Pectis brevipedunculata and its main constituent citral in rat aorta.
Vasodilator activity of the essential oil from aerial parts of Pectis brevipedunculata and its main constituent citral in rat aorta.
Molecules (Basel, Switzerland) (2013-03-09)
Sharlene Lopes Pereira, André Mesquita Marques, Roberto Takashi Sudo, Maria Auxiliadora Coelho Kaplan, Gisele Zapata-Sudo
ZUSAMMENFASSUNG
The essential oil of Pectis brevipedunculata (EOPB), a Brazilian ornamental aromatic grass, is characterized by its high content of citral (81.9%: neral 32.7% and geranial 49.2%), limonene (4.7%) and α-pinene (3.4%). Vasodilation induced by EOPB and isolated citral was investigated in pre-contracted vascular smooth muscle, using thoracic aorta from Wistar Kyoto (WKY) rats which was prepared for isometric tension recording. EOPB promoted intense relaxation of endothelium-intact and denuded aortic rings with the concentration to induce 50% of the maximal relaxation (IC50) of 0.044% ± 0.006% and 0.093% ± 0.015% (p < 0.05), respectively. The IC50 values for citral in endothelium-intact and denuded rings were 0.024% ± 0.004% and 0.021% ± 0.004%, respectively (p > 0.05). In endothelium-intact aorta, EOPB-induced vasorelaxation was significantly reduced by L-NAME, a nitric oxide synthase inhibitor. The vasodilator activity of citral was increased in the KCl-contracted aorta and citral attenuated the contracture elicited by Ca2+ in depolarized aorta. EOPB and citral elicited vasorelaxation on thoracic aorta by affecting the NO/cyclic GMP pathway and the calcium influx through voltage-dependent L-type Ca2+ channels, respectively.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Citral, 95%
Sigma-Aldrich
Citral, natural, ≥96%, FCC, FG
Sigma-Aldrich
Citral, mixture of cis and trans, ≥96%, FG