Merck
  • Application of a europium complex, Eu(AA)3phen (AA=acrylic acid, phen=1,10-phenanthroline) as a spectroscopic probe and cleaving reagent of DNA.
Application of a europium complex, Eu(AA)3phen (AA=acrylic acid, phen=1,10-phenanthroline) as a spectroscopic probe and cleaving reagent of DNA.
Materials science & engineering. C, Materials for biological applications (2013-03-19)
Ling Li, Quan Pan, Gong Wu Song
ZUSAMMENFASSUNG
A complex Eu(AA)3phen was synthesized and characterized by elemental analysis, IR spectroscopy and (1)H NMR. Interaction between Eu(AA)3phen and DNA was studied by UV/visible absorption spectroscopy, fluorescence spectrophotometer, circular-dichroism (CD) spectra and gel-electrophoresis measurements. Absorption spectral indicated that Eu(AA)3phen binding to DNA was an electrostatic mode, which was authenticated by the effect of ionic strength, thermal denaturation and fluorescence quenching experiments. The intrinsic binding constant Kb of Eu(AA)3phen was calculated to be 1.6×10(4) L mol(-1).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acrylsäure, anhydrous, contains 200 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
1,10-Phenanthrolin, ≥99%
Sigma-Aldrich
Zinkacrylat, 98%