Merck
  • Optimization of a lateral flow immunoassay for the ultrasensitive detection of aflatoxin M1 in milk.
Optimization of a lateral flow immunoassay for the ultrasensitive detection of aflatoxin M1 in milk.
Analytica chimica acta (2013-04-02)
Laura Anfossi, Claudio Baggiani, Cristina Giovannoli, Flavia Biagioli, Gilda D'Arco, Gianfranco Giraudi
ZUSAMMENFASSUNG
A high sensitive immunoassay-based lateral flow device for semi-quantitatively determine aflatoxin M1 (AFM1) in milk was developed. Investigation and optimization of the competitor design and of the gold-labelling strategy allowed the attainment of the ultra-sensitive assessment of AFM1 contamination at nanograms per litre level (LOD 20 ng L(-1), IC50 99 ng L(-1)), as requested by European regulations. A one order of magnitude detectability enhancement in comparison to previously reported gold colloid immunochromatographic assays for this toxin was obtained. Direct detection of the target toxin in milk could be obtained by acquiring images of the strips and correlating intensities of the coloured lines with analyte concentrations. The one-step assay can be completed in 17 min, including a very simple and rapid sample preparation, which allowed the application of the assay to milk samples which differ in fat and protein contents. Although imprecise (mean RSD about 30%), the method proved to be accurate and sensitive enough to allow the correct attribution of sample as compliant or non-compliant according to EU legislation in force. Agreeing results to those of a reference ELISA were obtained on 40 milk samples by matrix-matched calibration in pasteurized milk.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aflatoxin M1, from Aspergillus flavus
Supelco
Aflatoxin M1 -Lösung, 0.5 μg/mL in acetonitrile, analytical standard