Merck
  • The usefulness of three-dimensional cell culture in induction of cancer stem cells from esophageal squamous cell carcinoma cell lines.
The usefulness of three-dimensional cell culture in induction of cancer stem cells from esophageal squamous cell carcinoma cell lines.
Biochemical and biophysical research communications (2013-04-23)
Daisuke Fujiwara, Kazunori Kato, Shigeo Nohara, Yoshimi Iwanuma, Yoshiaki Kajiyama
ZUSAMMENFASSUNG
In recent years, research on resistance to chemotherapy and radiotherapy in cancer treatment has come under the spotlight, and researchers have also begun investigating the relationship between resistance and cancer stem cells. Cancer stem cells are assumed to be present in esophageal cancer, but experimental methods for identification and culture of these cells have not yet been established. To solve this problem, we created spheroids using a NanoCulture® Plate (NCP) for 3-dimensional (3-D) cell culture, which was designed as a means for experimentally reproducing the 3-D structures found in the body. We investigated the potential for induction of cancer stem cells from esophageal cancer cells. Using flow cytometry we analyzed the expression of surface antigen markers CD44, CD133, CD338 (ABCG2), CD318 (CDCP1), and CD326 (EpCAM), which are known cancer stem cell markers. None of these surface antigen markers showed enhanced expression in 3-D cultured cells. We then analyzed aldehyde dehydrogenase (ALDH) enzymatic activity using the ALDEFLUOR reagent, which can identify immature cells such as stem cells and precursor cells. 3-D-cultured cells were strongly positive for ALDH enzyme activity. We also analyzed the expression of the stem cell-related genes Sox-2, Nanog, Oct3/4, and Lin28 using RT-PCR. Expression of Sox-2, Nanog, and Lin28 was enhanced. Analysis of expression of the hypoxic surface antigen marker carbonic anhydrase-9 (CA-9), which is an indicator of cancer stem cell induction and maintenance, revealed that CA-9 expression was enhanced, suggesting that hypoxia had been induced. Comparison of cancer drug resistance using cisplatin and doxorubicin in 3-D-cultured esophageal cancer cells showed that cancer drug resistance had increased. These results indicate that 3-D culture of esophageal squamous cell carcinoma lines is a useful method for inducing cancer stem cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Carboanhydrase aus Rindererythrocyten, lyophilized powder, ≥2,000 W-A units/mg protein
Sigma-Aldrich
Carboanhydrase aus Rindererythrocyten, ≥95% (SDS-PAGE), specific activity ≥3,500 W-A units/mg protein, lyophilized powder
Sigma-Aldrich
Carboanhydrase aus Rindererythrocyten, BioReagent, suitable for GFC marker
Sigma-Aldrich
Karbonanhydraseisozym II aus Rindererythrocyten, lyophilized powder, ≥3,000 W-A units/mg protein
Sigma-Aldrich
Carboanhydrase aus Rindererythrocyten, For use as a marker in SDS-PAGE
Sigma-Aldrich
Kohlensäureanhydrase I aus Humanerythrocyten
Sigma-Aldrich
Carboanhydrase aus Rindererythrocyten, non-denaturing PAGE marker
Sigma-Aldrich
Carbonic Anhydrase Isozyme II human, ≥80%, powder, ≥3,000 W-A units/mg protein
Sigma-Aldrich
Karbonanhydraseisozym II aus Rindererythrocyten, lyophilized powder, ≥2,000 W-A units/mg protein
Supelco
Kohlensäureanhydrase I aus Humanerythrocyten, Isoelectric focusing marker, pI 6.6
Sigma-Aldrich
Karbonanhydraseisozym II aus Rindererythrocyten, Isoelectric focusing marker, pI 5.4