Merck
Identification and determination of selected histamine antagonists by densitometric method.
Acta poloniae pharmaceutica (2013-04-25)
Krystyna Czerwińska, Elzbieta Wyszomirska, Aleksander P Mazurek
ZUSAMMENFASSUNG
The conditions for identification were determined for four histamine antagonists: clemastine fumarate, loratadine, cetirizine dihydrochloride and desloratadine by TLC (thin-layer chromatography) method. The selected chromatographic conditions were used to develop a densitometric method for the content determination of the histamine antagonists in medicinal products and substances. The statistical data showed adequate accuracy and precision of the developed methods.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cetirizin -dihydrochlorid, ≥98% (HPLC)
Sigma-Aldrich
Loratadin, ≥98% (HPLC), powder
Sigma-Aldrich
Clemastine fumarate salt, ≥98% (HPLC)
Sigma-Aldrich
Cetirizin -dihydrochlorid, ≥98.0% (HPLC)
Cetirizin -dihydrochlorid, European Pharmacopoeia (EP) Reference Standard
Loratadin, European Pharmacopoeia (EP) Reference Standard
Cetirizin für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Loratadin für die Systemeignung, European Pharmacopoeia (EP) Reference Standard