Merck
  • Thalamic glutamate/glutamine in restless legs syndrome: increased and related to disturbed sleep.
Thalamic glutamate/glutamine in restless legs syndrome: increased and related to disturbed sleep.
Neurology (2013-04-30)
Richard P Allen, Peter B Barker, Alena Horská, Christopher J Earley
ZUSAMMENFASSUNG
To evaluate possible abnormal increase in thalamic glutamate/glutamine levels for restless legs syndrome (RLS) indicating increased glutamatergic activity producing arousal that at night disrupts and shortens sleep. (1)H MRS of the right thalamus was performed using a 1.5 T GE MRI scanner and the PROBE-P (PRESS) on 28 patients with RLS and 20 matched controls. The Glx signal (combination of mostly glutamate [Glu] and glutamine [Gln]) was assessed as a ratio to the total creatine (Cr). This study tested 2 primary hypotheses: 1) higher thalamic Glx/Cr for patients with RLS than controls; 2) thalamic Glx/Cr correlates with increased wake during the sleep period. The Glx/Cr was higher for patients with RLS than controls (mean ± SD 1.20 ± 0.73 vs 0.80 ± 0.39, t = 2.2, p = 0.016) and correlated significantly with the wake time during the sleep period (r = 0.61, p = 0.007) and all other RLS-related polysomnographic sleep variables (p < 0.05) except for periodic leg movements during sleep (PLMS)/hour. The primary findings introduce 2 new related dimensions to RLS: abnormalities in a major nondopaminergic neurologic system and the arousal disturbance of sleep. The strong relation of the arousal sleep disturbance to glutamate and the lack of relation to the PLMS motor features of RLS contrasts with the reverse for dopamine of a limited relation to arousal sleep disturbance but strong relation to PLMS. Understanding this dichotomy and the interaction of these 2 differing systems may be important for understanding RLS neurobiology and developing better treatments for RLS.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Glutamin -Lösung, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
SAFC
L-Glutaminlösung 200 mmol/l, 29.23 mg/mL in saline, solution, suitable for cell culture
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
SAFC
L-Glutamin
Sigma-Aldrich
Creatin, anhydrous
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Sigma-Aldrich
L-Glutamin -Lösung, 200 mM, Hybri-Max, sterile-filtered, suitable for hybridoma
Supelco
L-Glutamin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland