Merck
Deacetylation of p53 induces autophagy by suppressing Bmf expression.
The Journal of cell biology (2013-05-01)
Amelia U Contreras, Yohannes Mebratu, Monica Delgado, Gilbert Montano, Chien-An A Hu, Stefan W Ryter, Augustine M K Choi, Yuting Lin, Jialing Xiang, Hitendra Chand, Yohannes Tesfaigzi
ZUSAMMENFASSUNG
Interferon γ (IFN-γ)-induced cell death is mediated by the BH3-only domain protein, Bik, in a p53-independent manner. However, the effect of IFN-γ on p53 and how this affects autophagy have not been reported. The present study demonstrates that IFN-γ down-regulated expression of the BH3 domain-only protein, Bmf, in human and mouse airway epithelial cells in a p53-dependent manner. p53 also suppressed Bmf expression in response to other cell death-stimulating agents, including ultraviolet radiation and histone deacetylase inhibitors. IFN-γ did not affect Bmf messenger RNA half-life but increased nuclear p53 levels and the interaction of p53 with the Bmf promoter. IFN-γ-induced interaction of HDAC1 and p53 resulted in the deacetylation of p53 and suppression of Bmf expression independent of p53's proline-rich domain. Suppression of Bmf facilitated IFN-γ-induced autophagy by reducing the interaction of Beclin-1 and Bcl-2. Furthermore, autophagy was prominent in cultured bmf(-/-) but not in bmf(+/+) cells. Collectively, these observations show that deacetylation of p53 suppresses Bmf expression and facilitates autophagy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Buttersäure, ≥99%
Supelco
Buttersäure, analytical standard
Sigma-Aldrich
Buttersäure, ≥99%, FG
Sigma-Aldrich
Buttersäure, natural, ≥99%, FCC, FG
Sigma-Aldrich
Anti-Acetyl-Histon-H4-Antikörper, 1 mg/mL, Upstate®
Sigma-Aldrich
Anti-HDAC1-Antikörper, Upstate®, from rabbit