Merck
Effect of polishing and finishing procedures on the surface integrity of restorative ceramics.
American journal of dentistry (2013-06-04)
Tetsurou Odatsu, Ryo Jimbo, Ann Wennerberg, Ikuya Watanabe, Takashi Sawase
ZUSAMMENFASSUNG
To investigate the effect of surface polishing and finishing methods on the surface roughness of restorative ceramics. Disk specimens were prepared from feldspar-based, lithium disilicate-based, fluorapatite leucite-based and zirconia ceramics. Four kinds of surface polishing/finishing methods evaluated were: Group 1: carborundum points (CP); Group 2: silicon points (SP); Group 3: diamond paste (DP); Group 4: glazing (GZ). Surface roughness was measured using an interferometer and the parameters of Sa (average height deviation of the surface) and St (maximum peak-to-valley height of the surface) were evaluated. Data were statistically analyzed using two-way ANOVA (P < 0.05) followed by post-hoc test. The mean values were also compared by Student's t-test. Specimen surfaces were evaluated by 3-D images using an interferometer. The zirconia showed the least surface roughness (Sa and St) values after grinding with carborundum points. The significantly lowest Sa values and St values were obtained for lithium disilicate and zirconia ceramics surfaces finished with DP and GZ. The fluorapatite leucite ceramic showed significantly reduced Sa and St values from DP to GZ. The feldspathic porcelain showed the highest surface roughness values among all types of ceramics after all of the polishing/finishing procedures.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Zirkonium(IV)-oxid, powder, 5 μm, 99% trace metals basis
Sigma-Aldrich
Silizium, powder, −325 mesh, 99% trace metals basis
Sigma-Aldrich
Zirkonium(IV)-oxid, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Zirconium, powder, −100 mesh
Sigma-Aldrich
Silizium, wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silizium, nanopowder, <100 nm particle size (TEM), ≥98% trace metals basis
Sigma-Aldrich
Silizium, wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Diamant, nanopowder, <10 nm particle size (TEM), ≥97% trace metals basis
Sigma-Aldrich
Silizium, wafer (single side polished), <100>, P-type, contains boron as dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Zirkonium(IV)-oxid, 99.99% trace metals basis (purity excludes ~2% HfO2)
Sigma-Aldrich
Silizium, wafer (single side polished), <100>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Zirconium, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Zirkonium(IV)-oxid -Lösung, nanoparticles, dispersion, <100 nm particle size (BET), 5 wt. % in H2O
Sigma-Aldrich
Zirconium, rod, diam. 6.35 mm, ≥99% trace metals basis
Sigma-Aldrich
Silizium, wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silizium, wafer (single side polished), contains phosphorus as dopant, <111>, N-type, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silizium, pieces, 99.95% trace metals basis
Sigma-Aldrich
Zirconium, sponge, ≥99% trace metals basis
Sigma-Aldrich
Silizium, wafer, <111>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.3 mm
Sigma-Aldrich
Silizium, wafer (single side polished), <100>, N-type, contains phosphorus as dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Diamant, nanopowder, <10 nm particle size (TEM), ≥95% trace metals basis
Sigma-Aldrich
Zirkonium(IV)-oxid -Lösung, nanoparticles, dispersion, <100 nm particle size (BET), 10 wt. % in H2O
Sigma-Aldrich
Silizium, wafer (single side polished), <100>, N-type, contains phosphorus as dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Diamant, synthetic monocrystalline powder, ≤1 μm
Sigma-Aldrich
Silizium, wafer (single side polished), <111>, P-type, contains boron as dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silizium, wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm