Merck
  • The DMAP interaction domain of UDP-GlcNAc:lysosomal enzyme N-acetylglucosamine-1-phosphotransferase is a substrate recognition module.
The DMAP interaction domain of UDP-GlcNAc:lysosomal enzyme N-acetylglucosamine-1-phosphotransferase is a substrate recognition module.
Proceedings of the National Academy of Sciences of the United States of America (2013-06-05)
Yi Qian, Heather Flanagan-Steet, Eline van Meel, Richard Steet, Stuart A Kornfeld
ZUSAMMENFASSUNG
UDP-GlcNAc:lysosomal enzyme N-acetylglucosamine-1-phosphotransferase (GlcNAc-1-phosphotransferase) is an α2β2γ2 heterohexamer that mediates the initial step in the formation of the mannose 6-phosphate recognition signal on lysosomal acid hydrolases. We previously reported that the specificity of the reaction is determined by the ability of the α/β subunits to recognize a conformation-dependent protein determinant present on the acid hydrolases. We now present evidence that the DNA methyltransferase-associated protein (DMAP) interaction domain of the α subunit functions in this recognition process. First, GST-DMAP pulled down several acid hydrolases, but not nonlysosomal glycoproteins. Second, recombinant GlcNAc-1-phosphotransferase containing a missense mutation in the DMAP interaction domain (Lys732Asn) identified in a patient with mucolipidosis II exhibited full activity toward the simple sugar α-methyl d-mannoside but impaired phosphorylation of acid hydrolases. Finally, unlike the WT enzyme, expression of the K732N mutant in a zebrafish model of mucolipidosis II failed to correct the phenotypic abnormalities. These results indicate that the DMAP interaction domain of the α subunit functions in the selective recognition of acid hydrolase substrates and provides an explanation for the impaired phosphorylation of acid hydrolases in a patient with mucolipidosis II.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Acetyl-D-Glucosamin, ≥99% (HPLC)
Sigma-Aldrich
N-Acetyl-D-Glucosamin, suitable for cell culture, BioReagent
Sigma-Aldrich
N-Acetyl-D-Glucosamin, ≥95% (HPLC)
Sigma-Aldrich
D-Mannose-6-Phosphat Natriumsalz, ≥98% (HPLC)
Sigma-Aldrich
D-Mannose-6-Phosphat Dinatriumsalz Hydrat, ≥97.0% dry basis (enzymatic)
Sigma-Aldrich
N,N′-Bis(2-aminoethyl)-1,3-propandiamin, 97%