Merck
  • Antidiabetic activity of heart wood of Pterocarpus marsupium Roxb. and analysis of phytoconstituents.
Antidiabetic activity of heart wood of Pterocarpus marsupium Roxb. and analysis of phytoconstituents.
Indian journal of experimental biology (2013-07-05)
Akansha Mishra, Rohit Srivastava, Swayam Prakash Srivastava, Sudeep Gautam, Akhilesh Kumar Tamrakar, Rakesh Maurya, Arvind K Srivastava
ZUSAMMENFASSUNG
The crude powder, ethanolic extract and aqueous, chloroform, hexane and n-butanol soluble fractions of ethanolic extract of heart wood of P. marsupium showed marked improvement on oral glucose tolerance post sucrose load in normal rats. All these fractions except aqueous fraction showed improvement on oral glucose tolerance post sucrose load on streptozotocin (STZ)-induced diabetic rats. The crude powder, ethanolic extract and hexane and n-butanol fractions showed marked decline in blood glucose level on STZ-induced diabetic rats. The ethanolic extract (100 mg/kg body weight) when given to STZ-induced diabetic rats for 10 consecutive days declined blood glucose, improved OGTT and increased their serum insulin levels. The ethanolic extract also showed marked improvement on oral glucose tolerance on high fat-low dosed STZ-induced diabetic rats and neonatally STZ treated rats. The ethanolic extract of P. marsupium also showed marked antidyslipidemic effects on high fat diet fed Syrian golden hamsters. Altered renal and hepatic function markers and serum insulin levels of high fat diet fed-low dosed STZ-treated diabetic rats were also found towards normalization when these animals were treated with ethanolic extract of P. marsupium for 28 consecutive days. The four out of five phenolic C-glycosides isolated from n-butanol fraction of ethanolic extract of P. marsupium enhanced glucose uptake by skeletal muscle cells (C2C12) in a dose dependent manner. It may primarily be concluded that phenolic-C-glycosides present in P. marsupium heart wood are the phytoconstituents responsible for the antihyperglycemic activity and validate the claim of antidiabetic activity of heart wood of P. marsupium.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Streptozocin, ≥75% α-anomer basis, ≥98% (HPLC), powder
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-100 (10 Ampullen/Kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Supelco
Ethanol -Lösung, certified reference material, 2000 μg/mL in methanol
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-200 (10 Ampullen/Kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol -Lösung, 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol -Lösung, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-20 (10 Ampullen/Kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol -Lösung, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®