Merck
  • The sequential use of washing and an electrochemical reduction process for the remediation of lead-contaminated soils.
The sequential use of washing and an electrochemical reduction process for the remediation of lead-contaminated soils.
Environmental technology (2013-07-11)
Aydeniz Demir, Nurcan Köleli
ZUSAMMENFASSUNG
A two-step method for the remediation of three different types of lead (Pb)-contaminated soil was evaluated. The first step included soil washing with ethylenediaminetetraacetic acid (EDTA) to remove Pb from soils. The washing experiments were performed with 0.05 M Na2EDTA at 1:10 soil to liquid ratio. Following the washing, Pb removal efficiency from soils ranged within 50-70%. After the soil washing process, Pb2+ ions in the washing solution were reduced electrochemically in a fixed-bed reactor. Lead removal efficiency with the electrochemical reduction at -2.0 V potential ranged within 57-76%. The overall results indicate that this two-step method is an environmentally-friendly and effective technology to remediate Pb-contaminated soils, as well as Pb-contaminated wastewater treatment due to the transformation of toxic Pb2+ ions into a non-hazardous metallic form (Pb(0)).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, suitable for electrophoresis, Molecular Biology, 99.0-101.0% (titration)
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, ACS reagent, 99.0-101.0%
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, Molecular Biology, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Supelco
Dinatriumedetat Dihydrat, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, Sigma Grade, suitable for plant cell culture, 98.5-101.5%
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
Sigma-Aldrich
Ethylendiamintetraessigsäure Calcium-Dinatriumsalz Hydrat
Sigma-Aldrich
Blei, powder, −100 mesh, 99.95% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, 2%, solution
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, BioUltra, 98.5-101.5%
Sigma-Aldrich
Ethylendiamintetraessigsäure, ≥98.0% (KT)
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, ≥97.0% (KT)
Sigma-Aldrich
Blei, shot, <2 mm, 99.9% trace metals basis
Sigma-Aldrich
Blei, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Blei, shot, 1-3 mm, 99.995% trace metals basis