Merck
  • Facile decoration of Au nanoparticles on CdS nanorods by polyoxometalate with enhanced photocatalytic activities toward hydrogen evolution.
Facile decoration of Au nanoparticles on CdS nanorods by polyoxometalate with enhanced photocatalytic activities toward hydrogen evolution.
Journal of nanoscience and nanotechnology (2013-08-02)
Xiaolin Xing, Rongji Liu, Zhanli Wang, Baozeng Ren, Zhenxi Jiang, Hui Zhao, Hongbin Cao, Guangjin Zhang, Tao Zhang
ZUSAMMENFASSUNG
The tri-component hybrid CdS nanorods (NRs)/Au nanoparticles (NPs)@polyoxometalate (POM) was successfully prepared by a facile, efficient and green method. The structural properties and component analysis were studied by Transmission electron microscopy (TEM), X-ray Diffraction (XRD) and UV-Vis spectra. The POMs sever as not only reductant and bridge molecules, but also as co-catalyst to play an important role in the photocatalytic process. The as-prepared nanohybrid shows obviously enhanced photocatalytic activity toward photocatalytic evolution of hydrogen.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasserstoff, ≥99.99%
Sigma-Aldrich
Gold, rod, diam. 3.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, powder, <45 μm, 99.99% trace metals basis
Sigma-Aldrich
Gold, nanopowder, <100 nm particle size, 99.9% trace metals basis
Sigma-Aldrich
Gold, powder, <10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Goldbeschichteter Glasobjektträger, layer thickness 100 Å, 99.999% (Au)
Sigma-Aldrich
Gold, wire, diam. 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, beads, 1-6 mm, 99.999% trace metals basis
Sigma-Aldrich
Cadmiumselenid, -325 Mesh particle size, 99.99% trace metals basis, electronic grade
Sigma-Aldrich
Goldbeschichteter Glasobjektträger, layer thickness 1000 Å, 99.999% (Au)
Sigma-Aldrich
Gold, foil, thickness 0.1 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.05 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.1 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, evaporation slug, diam. × L 0.3 cm × 0.6 cm, 99.99% trace metals basis
Sigma-Aldrich
Gold, powder, <850 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 1.0 mm, 99.997% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.025 mm, 99.99% trace metals basis
Sigma-Aldrich
Goldbeschichtete Glasabdeckfolie, L × W × thickness 22 mm × 22 mm × 130-170 μm, square, layer thickness 100 Å, 99.999% (Au)
Sigma-Aldrich
Gold, wire, diam. 0.5 mm, 99.999% trace metals basis
Sigma-Aldrich
Gold, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.99% trace metals basis
Sigma-Aldrich
Goldbeschichtete Glasabdeckfolie, 99.999% (Au), diam. × thickness 15 mm × 130-170 μm, layer thickness 100 Å
Sigma-Aldrich
Gold, wire, diam. 0.127 mm, 99.99% trace metals basis