Merck
  • Pre-oxidation with KMnO4 changes extra-cellular organic matter's secretion characteristics to improve algal removal by coagulation with a low dosage of polyaluminium chloride.
Pre-oxidation with KMnO4 changes extra-cellular organic matter's secretion characteristics to improve algal removal by coagulation with a low dosage of polyaluminium chloride.
Journal of environmental sciences (China) (2013-08-08)
Lei Wang, Junlian Qiao, Yinghui Hu, Lei Wang, Long Zhang, Qiaoli Zhou, Naiyun Gao
ZUSAMMENFASSUNG
Microcystis aeruginosa was used to study the effect of KMnO4 pre-oxidation on algal removal through coagulation with polyaluminium chloride (PAC). KMnO4 pre-oxidation improved the coagulation efficiency of algal at a low dosage of PAC. The optimal KMnO4 feeding period was in the stationary growth phase of Microcystis aeruginosa. KMnO4 traumatized the algal cells and stimulated cellular release of organic matter, contributing to the pool of extra-cellular organic matter (EOM). KMnO4 also decomposed EOM, especially small molecular weight EOM. Lower concentrations of KMnO4, such as 2 mg/L, induced algae cells to produce moderate amounts of new EOM with molecular weights of 11, 280, and 1500 kDa. These relatively large molecules combined easily with PAC, promoting coagulation and removal of algae. High concentrations of KMnO4 lysed algae cells and produced much high-molecular-weight EOM that did not enhance flocculation by PAC at lower dosages.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumpermanganat, ACS reagent, ≥99.0%
Sigma-Aldrich
Aktivkohle, DARCO®, −100 mesh particle size, powder
Sigma-Aldrich
Aktivkohle, powder, -100 particle size (mesh), decolorizing
Sigma-Aldrich
Aluminiumhydroxid, reagent grade
Sigma-Aldrich
Kaliumpermanganat, ACS reagent, ≥99.0%, low in mercury
Sigma-Aldrich
Aktivkohle, DARCO®, 20-40 mesh particle size, granular
Sigma-Aldrich
Aktivkohle-Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Aktivkohle, untreated, granular, ≤5 mm
Sigma-Aldrich
Aktivkohle, DARCO®, 4-12 mesh particle size, granular
Sigma-Aldrich
Aktivkohle, DARCO®, 12-20 mesh, granular
Sigma-Aldrich
Kaliumpermanganat, 97%
Sigma-Aldrich
Kohlenstoff, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Supelco
Aktivkohle, powder
Sigma-Aldrich
Aktivkohle, acid-washed with hydrochloric acid
Sigma-Aldrich
Kohlenstoff, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Aktivkohle-Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Sigma-Aldrich
Aktivkohle, meets USP testing specifications
Supelco
Aktivkohle, puriss. p.a., powder
Sigma-Aldrich
Aktivkohle-Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Aktivkohle, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Aktivkohle-Norit®, Norit® RB3, steam activated, rod
Sigma-Aldrich
Aktivkohle-Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Aktivkohle, untreated, granular, 20-60 mesh
Supelco
Aktivkohle-Norit®, Norit® RBAA-3, rod
Millipore
Aktivkohle, suitable for GC
Supelco
Aktivkohle, for the determination of AOX, 50-150 μm particle size
Sigma-Aldrich
Kaliumpermanganat, meets USP testing specifications