Merck
Influences of perfluorooctanoic acid on the aggregation of multi-walled carbon nanotubes.
Journal of environmental sciences (China) (2013-08-08)
Chengliang Li, Andreas Schäffer, Harry Vereecken, Marc Heggen, Rong Ji, Erwin Klumpp
ZUSAMMENFASSUNG
The aggregation of multi-walled carbon nanotubes (MWCNTs) in the aqueous phase not only inhibits their extensive utilization in various aspects but also dominates their environmental fate and transport. The role of surfactants at low concentration in the aggregation of MWCNTs has been studied, however the effect of perfluorinated surfactants at low concentration is uncertain. To understand this interfacial phenomenon, the influences of perfluorooctanoic acid (PFOA), and sodium dodecyl sulfate (SDS) as a control, on MWCNT aggregation in the aqueous phase were examined by the UV absorbency method. Influences of pH and cationic species on the critical coagulation concentration (CCC) value were evaluated. The CCC values were dependent on the concentration of PFOA, however a pronounced effect of SDS concentration on the CCC values was not observed. The CCC values of the MWCNTs were 51.6 mmol/L in NaCl and 0.28 mmol/L in CaCl2 solutions, which suggested pronounced differences in the effects of Na+ and Ca2+ ions on the aggregation of the MWCNTs. The presence of both PFOA and SDS significantly decreased the CCC values of the MWCNTs in NaCl solution. The aggregation of the MWCNTs took place under acidic conditions and was not notably altered under neutral and alkaline conditions due to the electrostatic repulsion of deprotonated functional groups on the surface of the MWCNTs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Calciumchlorid Dihydrat, ACS reagent, ≥99%
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Calciumchlorid Dihydrat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Perfluoroctansäure, 95%
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Calciumchlorid Dihydrat, Molecular Biology, ≥99.0%
Sigma-Aldrich
Calciumchlorid, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calciumchlorid Dihydrat, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
SAFC
Natriumchlorid -Lösung, 5 M
Supelco
Natriumdodecylsulfat, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Calciumchlorid, desiccant, ACS reagent, ≥96.0%
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Calciumchlorid, powder, 99.99% trace metals basis
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)