Merck
Optical methods for monitoring temperature in spectrophotometric analysers.
Annals of clinical biochemistry (1983-05-01)
T D O'Leary, J L Badenoch, R Bais
ZUSAMMENFASSUNG
A procedure is described for monitoring the temperature in the reaction cuvettes of analytical systems that use photometers. The method employs the temperature-dependent change in absorbance of solutions of either 3,5-dinitrosalicylic acid or cresol red. The procedure is simple to perform and is especially useful in monitoring the temperature in instruments such as centrifugal analysers where the reaction cuvette is inaccessible. In some of the instruments studied, methodological changes were required to ensure that reactions were carried out at the selected temperature.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3,5-Dinitrosalicylsäure, used in colorimetric determination of reducing sugars
Sigma-Aldrich
3,5-Dinitrosalicylsäure, 98%
Sigma-Aldrich
Kresolrot, indicator grade, Dye content 95 %
Sigma-Aldrich
Kresolrot Natriumsalz, indicator grade