Merck
The metabolic effect of dodecanedioic acid infusion in non-insulin-dependent diabetic patients.
Nutrition (Burbank, Los Angeles County, Calif.) (1998-05-20)
A V Greco, G Mingrone, E Capristo, G Benedetti, A De Gaetano, G Gasbarrini
ZUSAMMENFASSUNG
Dodecanedioic acid (C12) is an even-numbered dicarboxylic acid (DA). Dicarboxylic acids are water-soluble substances with a metabolic pathway intermediate to those of lipids and carbohydrates. Previous studies showed that contrary to other DAs, very low amounts of C12 are lost with urine. The effects of 46.6 mmol of C12 intravenous infusion for 195 min on blood glucose levels were investigated in five patients with non-insulin-dependent diabetes mellitus (NIDDM), with a good metabolic compensation, and in five healthy volunteers matched for gender, age, and body mass index. Blood samples were taken every 15 min for a period of 360 min to measure glucose, insulin, C-peptide, ketone bodies, and free fatty acid (FFA) levels, and 24-h urine samples were collected to measure C12 and urea excretion. Plasma and urinary C12 concentrations were determined by high-pressure liquid chromatography (HPLC). Indirect calorimetry was continuously performed both basally and during the study period. The average 24-h urinary excretion of C12 was 6.5% versus 6.7% of the administered dose, respectively, in NIDDM patients and in healthy controls. The area under the curve (AUC) values of plasma C12 were 279.9 +/- 42.7 mumol in NIDDM patients and 219.7 +/- 14.0 mumol in controls (P = ns). Plasma glucose levels significantly decreased in NIDDM patients during C12 infusion (from 7.8 +/- 0.6 to 5.4 +/- 0.8 mM at the end of the study period, P < 0.05). Lactate plasma concentration decreased in NIDDM patients from 3.5 +/- 0.2 to 1.5 +/- 0.1 mM (P < 0.001), whereas blood pyruvate increased at the end of the experimental session from 26.0 +/- 11.6 to 99.5 +/- 14.9 microM (P < 0.01). Free fatty acids decreased in diabetic patients from the beginning until the end of C12 infusion, although this difference did not reach statistical significance. No significant increase was found between basal and final values in VO2 consumption and in the values of nonprotein respiratory quotient in both groups of subjects examined. The experimental data indicate that C12 infusion decreases plasma glucose levels in NIDDM patients to normal range without influencing plasma insulin levels. The balance between pyruvate and lactate was affected by C12 infusion only in diabetics patients. C12 might represent a fuel substrate immediately available for tissue energy requirements, especially in conditions such as diabetes mellitus in which glucose metabolism is impaired.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Aceton, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
1-Butanol, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Supelco
Aceton, analytical standard
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
Methylglyoxal -Lösung, ~40% in H2O
Sigma-Aldrich
Dimethylamin -Lösung, 40 wt. % in H2O
Sigma-Aldrich
Dimethylamin -Lösung, 2.0 M in THF
Sigma-Aldrich
Glycerin, FCC, FG
Sigma-Aldrich
3-Hydroxybuttersäure, 95%
Sigma-Aldrich
Dimethylamin, anhydrous, ≥99%
Sigma-Aldrich
Aceton, histological grade, ≥99.5%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Dodecandisäure, 99%
Sigma-Aldrich
1-Butanol, Molecular Biology, ≥99%
Supelco
Aceton, Pharmaceutical Secondary Standard; Certified Reference Material