Merck
  • Butein sensitizes human hepatoma cells to TRAIL-induced apoptosis via extracellular signal-regulated kinase/Sp1-dependent DR5 upregulation and NF-kappaB inactivation.
Butein sensitizes human hepatoma cells to TRAIL-induced apoptosis via extracellular signal-regulated kinase/Sp1-dependent DR5 upregulation and NF-kappaB inactivation.
Molecular cancer therapeutics (2010-06-03)
Dong-Oh Moon, Mun-Ock Kim, Yung Hyun Choi, Gi-Young Kim
ZUSAMMENFASSUNG
Tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) induces cell death in various types of cancer cells but has little or no effect on normal cells. Human hepatoma cells are resistant to TRAIL-induced apoptosis. Although butein is known to mediate anticancer, anti-inflammatory, and antioxidant activities, little is known about the mechanism of butein in terms of TRAIL-induced apoptosis of human hepatoma cells. In this study, we determined that butein enhances TRAIL-induced apoptosis in hepatoma cells through upregulation of DR5. Luciferase analysis showed that a 5'-flanking region containing four Sp1-binding sites within the DR5 promoter was enhanced by butein (-305/-300). Electrophoretic mobility shift assays and chromatin immunoprecipitation studies were used to analyze the elevation of Sp1 binding to DR5 promoter sites by butein. Point mutations of the Sp1-binding site also attenuated promoter activity. Furthermore, pretreatment of the blocking chimeric antibody and small interfering RNA for DR5 significantly suppressed TRAIL-mediated apoptosis by butein in Hep3B cells. Butein also stimulated extracellular signal-regulated kinase (ERK) activation, and the ERK inhibitor PD98059 blocked butein-induced DR5 expression and suppressed binding of Sp1 to the DR5 promoter. Additionally, generation of reactive oxygen species had no effect on cell viability, although pretreatment with N-acetyl-l-cysteine or glutathione inhibited combined treatment-induced reactive oxygen species. Indeed, butein repressed the TRAIL-mediated activation of NF-kappaB and decreased its transcriptional activity. Our results suggest that butein could sensitize certain human hepatoma cells to TRAIL-induced apoptosis through stimulating its death signaling and by repressing the survival function in these cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(R)-MG132
Sigma-Aldrich
PD 98059, PD 98059, CAS 167869-21-8, is a cell-permeable, selective & reversible inhibitor of MAP Kinase Kinase (MEK). Inhibits MAP Kinase activation and subsequent phosphorylation of MAP Kinase substrates.
Sigma-Aldrich
JNK-Inhibitor II, JNK Inhibitor II. SP600125, CAS 129-56-6, is a potent, cell-permeable, selective, and ATP competitive inhibitor of c-Jun N-terminal kinase (JNK; IC50 = 40 nM for JNK-1 & JNK-2 & 90 nM for JNK-3).
Sigma-Aldrich
Caspase-Inhibitor VI, Z-VAD-FMK, CAS 161401-82-7, is an irreversible pan caspase inhibitor. Does not require pretreatment with esterase for in vitro studies.