Merck
  • Differential effects of remacemide and desglycinyl-remacemide on epileptiform burst firing in the rat hippocampal slice.
Differential effects of remacemide and desglycinyl-remacemide on epileptiform burst firing in the rat hippocampal slice.
Neuroscience letters (2002-03-02)
Sarah Santangeli, Graeme J Sills, Trevor W Stone, Martin J Brodie
ZUSAMMENFASSUNG
Remacemide is a potential anticonvulsant drug with an active metabolite, desglycinyl-remacemide (DGR). Both moieties have been reported to block neuronal Na(+) channels and the N-methyl-D-aspartate (NMDA) subtype of glutamate receptor. The effects of remacemide and DGR on zero Mg(2+)/4-aminopyridine-induced epileptiform discharges were investigated in the rat hippocampal slice preparation and compared with carbamazepine (CBZ), a prototypic Na(+) channel blocker, and AR-R15896AR, a putative NMDA channel blocker. Remacemide (0-100 microM) was without significant effect, while DGR, CBZ and AR-R15896AR all decreased burst frequency in a concentration (0-100 microM) dependent manner. These findings suggest that remacemide is not sufficiently potent at the Na(+) channel or NMDA receptor to attenuate epileptiform activity in this model and that the anticonvulsant effects of the drug may be mediated by DGR.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Remacemide hydrochloride, ≥98% (HPLC)