Merck
  • The impacts of ozonation on oil sands process-affected water biodegradability and biofilm formation characteristics in bioreactors.
The impacts of ozonation on oil sands process-affected water biodegradability and biofilm formation characteristics in bioreactors.
Bioresource technology (2013-01-15)
Geelsu Hwang, Tao Dong, Md Sahinoor Islam, Zhiya Sheng, Leónidas A Pérez-Estrada, Yang Liu, Mohamed Gamal El-Din
ZUSAMMENFASSUNG
To examine the effects of the ozonation process (as an oxidation treatment for water and wastewater treatment applications) on microbial biofilm formation and biodegradability of organic compounds present in oil sands process-affected water (OSPW), biofilm reactors were operated continuously for 6weeks. Two types of biofilm substrate materials: polyethylene (PE) and polyvinylchloride (PVC), and two types of OSPW-fresh and ozonated OSPWs-were tested. Endogenous microorganisms, in OSPW, quickly formed biofilms in the reactors. Without ozonation, the bioreactor (using endogenous microorganisms) removed 13.8% of the total acid-extractable organics (TAO) and 18.5% of the parent naphthenic acids (NAs) from fresh OSPW. The combined ozonation and biodegradation process removed 87.2% of the OSPW TAO and over 99% of the OSPW parent NAs. Further UPLC/HRMS analysis showed that NA biodegradability decreased as the NA cyclization number increased. Microbial biofilm formation was found to depend on the biofilm substrate type.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Poly(vinylchlorid), low molecular weight
Sigma-Aldrich
Poly(vinylchlorid), high molecular weight
Sigma-Aldrich
Poly(vinylchlorid), average Mw ~233,000, average Mn ~99,000
Sigma-Aldrich
Naphthensäure, technical
Sigma-Aldrich
Poly(vinylchlorid), average Mw ~62,000, average Mn ~35,000
Supelco
Poly(vinylchlorid), Selectophore, high molecular weight
Supelco
Poly(vinylchlorid), analytical standard, average Mw 85,000 (Typical), average Mn 40,000 (Typical)
Poly(vinylchlorid), European Pharmacopoeia (EP) Reference Standard