Merck
  • In situ preparation of highly fluorescent pyrene-dyes from non-luminous precursors upon photoirradiation.
In situ preparation of highly fluorescent pyrene-dyes from non-luminous precursors upon photoirradiation.
Chemical communications (Cambridge, England) (2013-03-29)
Tatsuya Aotake, Hiroshi Tanimoto, Hidekatsu Hotta, Daiki Kuzuhara, Tetsuo Okujima, Hidemitsu Uno, Hiroko Yamada
ZUSAMMENFASSUNG
The non-luminous precursor, 2-(1-pyrenyl)-9,10-dihydro-9,10-ethanoanthracene-11,12-dione, was photochemically converted to highly-fluorescent 2-(1-pyrenyl)anthracene quantitatively in solution and in the PMMA film and the fluorescence quantum yield of the acene in benzonitrile was as high as 0.99.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pyren, 98%
Sigma-Aldrich
Pyren, puriss. p.a., suitable for fluorescence, ≥99.0% (GC)
Supelco
Pyren, analytical standard
Supelco
Pyren, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Pyren, sublimed grade, 99%
Pyren, BCR®, certified reference material