Merck
  • Efficient enzymatic hydrolysis of the bagasse pulp prepared with active oxygen and MgO-based solid alkali.
Efficient enzymatic hydrolysis of the bagasse pulp prepared with active oxygen and MgO-based solid alkali.
Carbohydrate polymers (2013-04-03)
Tujun Xie, Lu Lin, Chunsheng Pang, Junping Zhuang, Jianbin Shi, Qiulin Yang
ZUSAMMENFASSUNG
The enzymatic hydrolysis of the bagasse pulp prepared from the treatment process with active oxygen and MgO-based solid alkali was studied. The hydrolysates were tested by IC (ionic chromatography) for the analysis of monosaccharide. Additionally, the changes of pulp before and after hydrolysis were characterized with Fourier transform infrared spectroscopy (FTIR), scanning electron microscopy (SEM), X-ray diffraction (XRD), Kajaani cellulose automatic analyzer and atomic force microscopy (AFM) techniques. The results showed that an optimized sugar yield of 82.38% was obtained at the substrate concentration of 5% for 72h with the enzyme dosage of 15IU/g. Furthermore, as the length of the cellulose fiber decreased, the crystallinity of cellulose increased, and more depressions appeared on the surface of pulp after enzymatic hydrolysis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Magnesiumoxid, ACS reagent, 97%
Sigma-Aldrich
Cellulosepulver, microcrystalline, powder, 20 μm
Sigma-Aldrich
Magnesiumoxid, ≥99% trace metals basis, -325 mesh
Sigma-Aldrich
Cellulosepulver, microcrystalline, powder
Sigma-Aldrich
Cellulosepulver, fibers, (medium)
Sigma-Aldrich
Sigmacell Cellulose, Type 20, 20 μm
Supelco
Avicel® PH-101, ~50 μm particle size
Sigma-Aldrich
α-Cellulose, powder
Sigma-Aldrich
Magnesiumoxid, nanopowder, ≤50 nm particle size (BET)
Sigma-Aldrich
Magnesiumoxid, light, 95%
Sigma-Aldrich
Magnesiumoxid, ≥99.99% trace metals basis
Sigma-Aldrich
Cellulosepulver, colloidal, microcrystalline
Sigma-Aldrich
Magnesiumoxid, -10-+50 mesh, 98%
Sigma-Aldrich
Sigmacell Cellulose, Type 50, 50 μm
Sigma-Aldrich
Sigmacell Cellulose, Type 101, Highly purified, fibers
Sigma-Aldrich
Avicel® PH-101, tested according to Ph. Eur.
Sigma-Aldrich
α-Cellulose, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Magnesiumoxid, 99.99% trace metals basis
Supelco
Cellulosepulver, powder, suitable for column chromatography
Sigma-Aldrich
Magnesiumoxid, (single crystal substrate), ≥99.9% trace metals basis, <100>, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Magnesiumoxid, tested according to Ph. Eur., heavy
Supelco
Cellulosepulver, DS-0, powder, suitable for thin layer chromatography (TLC)
Sigma-Aldrich
Magnesiumoxid, BioUltra, ≥97.0% (calcined substance, KT)
Supelco
Cellulosepulver, acid washed, powder, suitable for column chromatography
Supelco
Cellulosepulver, DFS-0, microcrystalline, suitable for thin layer chromatography (TLC)
Supelco
Cellulosepulver, acid washed, from spruce, suitable for column chromatography