Merck
Anti-aging effects of co-enzyme Q10 on periodontal tissues.
Journal of dental research (2013-05-23)
T Yoneda, T Tomofuji, D Ekuni, T Azuma, Y Endo, K Kasuyama, T Machida, M Morita
ZUSAMMENFASSUNG
Oxidative stress is associated with age-related reactions. The anti-oxidative effects of a reduced form of co-enzyme Q10 (rCoQ10) suppress oxidative stress, which may contribute to the prevention of age-related inflammatory reactions. We examined the effects of topically applied rCoQ10 on periodontal inflammatory reactions in a rat aging model. Male Fischer 344 rats, 2 (n = 6) and 4 mos (n = 18) of age, were used. All of the two-month-old rats and 6 of the four-month-old rats were sacrificed and 12 remaining four-month-old rats received topically applied ointment with or without 1% rCoQ10 on the gingival surface until they reached 6 mos of age. The rats showed an age-dependent increase in circulating oxidative stress. RCoQ10 decreased oxidative DNA damage and tartrate-resistant acid-phosphatase-positive osteoclasts in the periodontal tissue at 6 mos of age as compared with the control. The same conditions lowered gene expression of caspase-1 and interleukin-1β in the periodontal tissue. Furthermore, Nod-like receptor protein 3 inflammasomes were less activated in periodontal tissues from rCoQ10-treated rats as compared with the control rats. Our results suggest that rCoQ10 suppresses age-related inflammatory reactions and osteoclast differentiation by inhibiting oxidative stress.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phosphatase, sauer aus Kartoffeln, lyophilized powder, ≥0.5 unit/mg solid
Sigma-Aldrich
Phosphatase, sauer aus Weizenkeimen, ≥0.4 unit/mg solid
Sigma-Aldrich
Phosphatase, sauer aus Kartoffeln, lyophilized powder, ≥3.0 units/mg solid
Sigma-Aldrich
Phosphatase, sauer aus Süsskartoffeln, ammonium sulfate suspension, ≥10.0 units/mg protein (modified Warburg-Christian)