Merck
[Pseudo-hyperkalemia and hyperleukocytosis].
Revue medicale de Bruxelles (2013-08-21)
A Goubella, A Thooft, P Antoine, J Creteur, J-L Vincent
ZUSAMMENFASSUNG
A pseudo-hyperkalemia may occur with hyperleukocytosis. It is important to recognize it early to avoid unnecessary or even dangerous treatment inducing hypokalemia. The pseudohyperkalemia is due to cell fragility coupled to mechanical phenomena during blood collecting. We report a case of pseudo-hyperkalemia in a context of acute myeloid leukemia.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Hydroxyharnstoff, 98%, powder
Sigma-Aldrich
Cytosin β-D-Arabinofuranosid, crystalline, ≥90% (HPLC)
Sigma-Aldrich
Cytosin β-D-Arabinofuranosid -hydrochlorid, crystalline