Merck
  • Analysis of volatile organic compounds in human urine by headspace gas chromatography-mass spectrometry with a multipurpose sampler.
Analysis of volatile organic compounds in human urine by headspace gas chromatography-mass spectrometry with a multipurpose sampler.
Journal of chromatography. A (1999-08-04)
H G Wahl, A Hoffmann, D Luft, H M Liebich
ZUSAMMENFASSUNG
A multipurpose sampler (Gerstel MPS), designed for liquid large volume, gaseous and headspace samples was used for the GC-MS analysis of organic volatiles in human urine. Headspace sampling with a volume-, temperature- and speed-controlled gas-tight syringe was combined with a temperature-controlled cold injection system (CIS) for cold trapping, enrichment and focusing of analytes. Regular 2-ml GC vials filled with 1 ml acidified urine were used as headspace sampling vials. A 100-vial autosampler tray was equipped with an additional temperature and heating time controlled "preheating station" for five vials. Profiles of organic volatiles in human urine were determined and 34 components identified. Trimethylamine (TMA) and 4-heptanone as two metabolites of medical interest were quantified. Calibration curves and intra assay imprecision for 4-heptanone concentrations in the range of 40 to 800 ng/ml showed a correlation coefficient of r = 0.9980 and a relative standard deviation (RSD) between 3.0 and 3.4%. Calibration curves and intra-assay imprecision for TMA concentrations in the range of medical interest from 0.5 to 20 micrograms/ml showed a correlation coefficient of r = 0.9968 and a RSD between 4.1 and 6.8%. The high practicability of the multipurpose sampler for both gaseous and liquid samples together with the here shown good reproducibility and sensitivity make this single CIS-GC-MS system very attractive for routine clinical use in metabolic profiling of organic volatiles (headspace) and non-volatiles (liquid).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Aceton, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
1-Butanol, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Supelco
Aceton, analytical standard
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
Methylglyoxal -Lösung, ~40% in H2O
Sigma-Aldrich
Dimethylamin -Lösung, 40 wt. % in H2O
Sigma-Aldrich
Dimethylamin -Lösung, 2.0 M in THF
Sigma-Aldrich
Glycerin, FCC, FG
Sigma-Aldrich
3-Hydroxybuttersäure, 95%
Sigma-Aldrich
Dimethylamin, anhydrous, ≥99%
Sigma-Aldrich
Aceton, histological grade, ≥99.5%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Dodecandisäure, 99%
Sigma-Aldrich
1-Butanol, Molecular Biology, ≥99%
Supelco
Aceton, Pharmaceutical Secondary Standard; Certified Reference Material