Merck
  • Dietary therapy in two patients with a mild form of sulphite oxidase deficiency. Evidence for clinical and biological improvement.
Dietary therapy in two patients with a mild form of sulphite oxidase deficiency. Evidence for clinical and biological improvement.
Journal of inherited metabolic disease (2000-02-22)
G Touati, E Rusthoven, E Depondt, C Dorche, M Duran, B Heron, D Rabier, M Russo, J M Saudubray
ZUSAMMENFASSUNG
We report an attempt at dietetic therapy in two unrelated patients with isolated sulphite oxidase deficiency, with a mild clinical course and late onset of symptoms. In case 1, disease started at 15 months with an acute crisis of agitation, unexplained crying and restlessness following otitis. Case 2 was diagnosed at 10 months when she presented with slight motor delay and dislocation of lenses. In both cases, sulphite oxidase activity measured in fibroblasts was undetectable. Therapy consisted of a diet low in protein from natural foods (daily methionine intake 130-150 mg) and a synthetic amino acid mixture (50 g per day) without cystine and methionine (Xmet, Cys Maxamaid, SHS International Ltd). A comparison of clinical and biochemical parameters was made between the period before treatment and after 2 years of treatment. Restriction in protein and sulphur amino acids brought about a dramatic decrease of urinary thiosulphate and S-sulphocysteine. It also brought about a generalized hypoaminoacidaemia with a low plasma methionine and cystine in both patients. Furthermore, both patients grew normally with no signs of neurological deterioration, and there was evidence of progress in psychomotor development.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Taurin, ≥99%
Sigma-Aldrich
Taurin, suitable for cell culture, meets USP testing specifications
Sigma-Aldrich
Taurin, BioUltra, ≥99.5% (T)
Supelco
Taurin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Cystein-S-Sulfat, ≥98% (TLC)
Sigma-Aldrich
Taurin, ≥98%, FG
SAFC
Taurin