Merck
Intravesical valrubicin in the treatment of carcinoma in situ of the bladder.
Expert opinion on pharmacotherapy (2001-10-05)
D D Kuznetsov, N F Alsikafi, R C O'Connor, G D Steinberg
ZUSAMMENFASSUNG
The propensity of patients with carcinoma in situ (CIS) of the bladder to progress to invasive and metastatic disease is clearly established. Today, the standard therapy in treating patients with CIS of the bladder is intravesical bacillus Calmette-Guerin (BCG). Nevertheless, patients who fail intravesical BCG have few viable options except to undergo a radical cystectomy. Valrubicin (N-trifluoroacetyladriamycin-14-valerate) is a new semisynthetic derivative of the anthracycline antibiotic doxorubicin that has been shown to benefit patients with BCG-refractory CIS of the bladder. Intravesical instillation of valrubicin is well-tolerated, safe and can be durable. Early non-randomised studies show promise and the current utilisation of this drug is limited to patients with BCG-refractory CIS of the bladder who are not good surgical candidates. Randomised studies of intravesical valrubicin for the treatment of superficial bladder cancer are ongoing.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Valeriansäure, ≥99%
Sigma-Aldrich
Valeriansäure, ≥99%, FCC, FG
Supelco
Valeriansäure, analytical standard