Merck
  • Metabolic effects of an enteral nutrition formula for diabetes: comparison with standard formulas in patients with type 1 diabetes.
Metabolic effects of an enteral nutrition formula for diabetes: comparison with standard formulas in patients with type 1 diabetes.
Clinical nutrition (Edinburgh, Scotland) (2003-09-27)
María del Carmen Crespillo, Gabriel Olveira, Maria Soledad Ruiz de Adana, Gemma Rojo-Martínez, Jorge García-Alemán, Pilar Olvera, Federico Soriguer, Antonio Muñoz
ZUSAMMENFASSUNG
To evaluate the metabolic response (glucose, cholesterol, triglycerides and beta-hydroxy-butyrate) in patients with type 1 diabetes after a trial breakfast with an enteral nutrition formula designed for patients with diabetes and compare it with standard formulas (with and without fibre). Each of 11 patients with type 1 diabetes consumed three types of liquid breakfast with a 1 week interval between each. (1). A standard diet (SD) with 49% carbohydrates, 35% lipids, 16% proteins-casein-and without fibre; (2). A fibre-enriched diet (FD): with 49% carbohydrates, 35% lipids, 16% casein and 15 g/1000 ml fibre; (3). A diet designed for patients with diabetes (DD) with 45% carbohydrates, 38% lipids, 16% soy protein and 15 g/1000 ml fibre. Each subject consumed 250 ml of each preparation at 9.00 AM after having administered their usual insulin dose, which was the same for each diet. Blood samples were taken at baseline and each 30 min, up to 150 min. The increase in postprandial glycaemia was lower with DD than with the standard preparations, reaching statistical significance at 60 min. There were no significant variations in the levels of cholesterol, triglycerides or beta-hydroxy-butyrate between the three preparations. After a trial breakfast, a diet designed for patients with diabetes provoked lower increases in postprandial glycaemia (with no changes in lipid or beta-hydroxy-butyrate levels) compared to the standard diets evaluated (with and without fibre).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Aceton, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
1-Butanol, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Supelco
Aceton, analytical standard
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
Methylglyoxal -Lösung, ~40% in H2O
Sigma-Aldrich
Dimethylamin -Lösung, 40 wt. % in H2O
Sigma-Aldrich
Dimethylamin -Lösung, 2.0 M in THF
Sigma-Aldrich
Glycerin, FCC, FG
Sigma-Aldrich
3-Hydroxybuttersäure, 95%
Sigma-Aldrich
Aceton, histological grade, ≥99.5%
Sigma-Aldrich
Dimethylamin, anhydrous, ≥99%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Dodecandisäure, 99%
Sigma-Aldrich
1-Butanol, Molecular Biology, ≥99%
Supelco
Aceton, Pharmaceutical Secondary Standard; Certified Reference Material