Merck
  • First ISNS Reference Preparation for Neonatal Screening for thyrotropin, phenylalanine and 17alpha-hydroxyprogesterone in blood spots.
First ISNS Reference Preparation for Neonatal Screening for thyrotropin, phenylalanine and 17alpha-hydroxyprogesterone in blood spots.
Journal of inherited metabolic disease (2007-06-19)
L H Elvers, J G Loeber, J-L Dhondt, M Fukushi, W H Hannon, T Torresani, D Webster
ZUSAMMENFASSUNG
Neonatal screening for congenital disorders like phenylketonuria (PKU), congenital hypothyroidism (CH) and congenital adrenal hyperplasia (CAH) is generally performed in dried blood spots on filter paper. The analytes of interest for testing for PKU, CH and CAH are phenylalanine, thyrotropin (TSH) and 17alpha-hydroxyprogesterone (17OHP), respectively. The International Society for Neonatal Screening (ISNS) decided to prepare a combined reference preparation for the three analytes on filter paper Schleicher & Schuell #903, Whatman BFC180 and Toyo Roshi 545. This 'First ISNS Reference Preparation for Neonatal Screening for TSH, phenylalanine and 17OHP in blood spots' (1st ISNS-RPNS) has been prepared by the RIVM (Bilthoven). The number of filter paper cards prepared, each with two sets of six blood spot calibrators, was 480, 42 and 69 for Schleicher & Schuell #903, Whatman BFC180 and Toyo Roshi 545, respectively. The volume of blood dispensed was 50 microl. The range of concentrations for TSH was 1-121 mIU/L blood, for phenylalanine 65-865 micromol/L blood and for 17OHP 2.2-302 nmol/L blood. The linearity of the blood spot calibrators and the homogeneity of the batch (only tested for Schleicher & Schuell) were good. The differences between the three filter papers were small: i.e. the potency of the ISNS-RPNS on Whatman and Toyo Roshi in terms of Schleicher & Schuell was between 0.98 and 1.09 for the three analytes. The 1st ISNS-RPNS for TSH, phenylalanine and 17OHP can be said to be suitable as formal reference preparation and as a source for (re)calibrating kit calibrators.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Leucin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Leucin, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Valin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Phenylalanin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Phenylessigsäure, 99%
Sigma-Aldrich
L-Phenylalanin, reagent grade, ≥98%
Sigma-Aldrich
L-Valin, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Mandelsäure, 99%
SAFC
L-Valin
Sigma-Aldrich
L-Phenylalanin, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
DL-3-Phenyl-milchsäure, ≥98%
Sigma-Aldrich
L-Valin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Phenylessigsäure, ≥99%, FCC, FG
Supelco
L-Phenylalanin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenylbrenztraubensäure, 98%
Sigma-Aldrich
L-Leucin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
17α-Hydroxyprogesteron, ≥95%
Supelco
L-Leucin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Leucin, 99%, FG
Supelco
L-Valin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-(−)-3-Phenylmilchsäure, 98%
Sigma-Aldrich
2-Hydroxyphenylessigsäure, ReagentPlus®, 99%
Supelco
17α-Hydroxyprogesterone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
3-Hydroxyphenylessigsäure, ≥99%
Sigma-Aldrich
L-Phenylalanin, 99%, FCC
Supelco
DL-Mandelsäure, analytical standard
Supelco
L-Phenylalanin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Leucin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Valin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland