Merck
  • Serum 1,5-anhydroglucitol (Glycomark) levels in children with and without type 1 diabetes mellitus.
Serum 1,5-anhydroglucitol (Glycomark) levels in children with and without type 1 diabetes mellitus.
Pediatric diabetes (2007-07-31)
Thanh M Nguyen, Luisa M Rodriguez, Kimberly J Mason, Rubina A Heptulla
ZUSAMMENFASSUNG
Postprandial hyperglycemia associated with diabetes is a risk factor for cardiovascular disease. Currently, glycated hemoglobin A(1c) (HgbA(1c)) and glycated protein fructosamine are not sensitive markers for acute and short-term hyperglycemia. 1,5-Anhydroglucitol (1,5-AG) (Glycomark; Tomen America, New York, NY, USA) is reported in adults with type 1 diabetes mellitus (T1DM) and type 2 diabetes mellitus (T2DM) as a marker for postmeal hyperglycemia. However, the reference ranges for 1,5-AG in normal children and children with T1DM are not known. We studied 1,5-AG levels in 10 control children (6 males and 4 females) and 10 children with T1DM (7 males and 3 females). The levels of 1,5-AG in the normal controls were higher than those in children with T1DM (24.60 +/- 3.99 microg/mL vs. 4.75 +/- 2.95 microg/mL; p < 0.0001). There were no gender differences noted. The 1,5-AG levels were negatively correlated with HgbA(1c) (r =-0.9366; p < 0.0001) and the peak postmeal plasma glucose concentrations (Pearson r =-7230; p = 0.0003). Our findings suggest that despite good glycemic control, postprandial glucose concentrations are elevated and that 1,5-AG showed a difference between controls and children with T1DM. The data are comparable with previous studies in normal adults and in those with T1DM and T2DM. They support the use of 1,5-AG concentrations, together with HgbA(1c), to evaluate therapy, especially to target postprandial hyperglycemia.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Aceton, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
1-Butanol, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Supelco
Aceton, analytical standard
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
Methylglyoxal -Lösung, ~40% in H2O
Sigma-Aldrich
Dimethylamin -Lösung, 40 wt. % in H2O
Sigma-Aldrich
Dimethylamin -Lösung, 2.0 M in THF
Sigma-Aldrich
Glycerin, FCC, FG
Sigma-Aldrich
3-Hydroxybuttersäure, 95%
Sigma-Aldrich
Dimethylamin, anhydrous, ≥99%
Sigma-Aldrich
Aceton, histological grade, ≥99.5%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Dodecandisäure, 99%
Sigma-Aldrich
1-Butanol, Molecular Biology, ≥99%
Supelco
Aceton, Pharmaceutical Secondary Standard; Certified Reference Material