Merck
  • Contribution of 3-hydroxyisobutyrate to the measurement of 3-hydroxybutyrate in human plasma: comparison of enzymatic and gas-liquid chromatography-mass spectrometry assays in normal and in diabetic subjects.
Contribution of 3-hydroxyisobutyrate to the measurement of 3-hydroxybutyrate in human plasma: comparison of enzymatic and gas-liquid chromatography-mass spectrometry assays in normal and in diabetic subjects.
Journal of lipid research (1989-11-01)
A Avogaro, D M Bier
ZUSAMMENFASSUNG
In this study we employed a capillary gas-liquid chromatographic-mass spectrometric (GLC-MS) method to measure the plasma concentrations of 3-hydroxybutyrate (3-OHB) and 3-hydroxyisobutyrate (3-OHIB) in overnight fasted diabetic subjects and in normal subjects. Plasma contents of 3-hydroxybutyrate measured in this fashion were identical to those obtained by enzymatic assay using a commercial preparation of beta-hydroxybutyrate dehydrogenase, indicating no significant contamination of this enzyme preparation with 3-hydroxyisobutyrate dehydrogenase. In normal individuals, plasma 3-OHIB concentration was 21 +/- 2 microM in the overnight fasted state and was higher in diabetic subjects (38 +/- 5 microM) and in subjects fasted for 72 h (97 +/- 4 microM). In the postabsorptive state, 3-OHIB was 33% the concentration of 3-OHB in normals and 17% that of 3-OHB in the diabetics.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Aceton, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
1-Butanol, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Supelco
Aceton, analytical standard
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
Methylglyoxal -Lösung, ~40% in H2O
Sigma-Aldrich
Dimethylamin -Lösung, 40 wt. % in H2O
Sigma-Aldrich
Dimethylamin -Lösung, 2.0 M in THF
Sigma-Aldrich
Glycerin, FCC, FG
Sigma-Aldrich
3-Hydroxybuttersäure, 95%
Sigma-Aldrich
Dimethylamin, anhydrous, ≥99%
Sigma-Aldrich
Aceton, histological grade, ≥99.5%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Dodecandisäure, 99%
Sigma-Aldrich
1-Butanol, Molecular Biology, ≥99%
Supelco
Aceton, Pharmaceutical Secondary Standard; Certified Reference Material