Merck
  • Plasma fatty acid pattern including diene-conjugated linoleic acid in ethanol users and patients with ethanol-related liver disease.
Plasma fatty acid pattern including diene-conjugated linoleic acid in ethanol users and patients with ethanol-related liver disease.
Alcoholism, clinical and experimental research (1986-12-01)
J Szebeni, C Eskelson, R Sampliner, B Hartmann, J Griffin, T Dormandy, R R Watson
ZUSAMMENFASSUNG
The level of conjugated dienes (CD) in the serum, conjugated linoleic acid isomer in the phospholipid fraction [18:2(9, 11)], and the fatty acid profile in both the serum and in the separated lipid classes were analyzed in current and previous alcohol abusers and in patients with alcoholic liver disease. None of the subjects consumed alcohol for at least 2-3 days prior to blood collection for analysis of lipids. There was no significant difference in CD or in the absolute level of 18:2(9, 11) among the groups, whereas the 18:2(9, 11)/18:2(9, 12) molar ratio was significantly elevated in patients with liver disease. Fatty acid analysis of the whole plasma extract showed that the absolute level of arachidonic acid increased from 4.8 +/- 3.5 mg/dl (n = 9) in lifelong abstainers to 13.4 +/- 4.0 (n = 8) in current ethanol abusers, and its relative level (related to the total fatty acids) from 1.73 +/- 1.34 to 4.55 +/- 1.01 in the same groups. An increased proportion of linoleic acid in the triglyceride and phospholipid fractions from current abusers and patients with liver disease was also found; the percentage of 18:2 in phospholipids increased from 5.4 +/- 5.0 (n = 8) in lifelong abstainers to 14.3 +/- 3.9 (n = 8) in current abusers and 12.7 +/- 2.2 (n = 10) in patients with liver disease. The percentage of 16:0 was significantly lower in the phospholipids of current abusers as compared to lifelong abstainers.(ABSTRACT TRUNCATED AT 250 WORDS)
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Harnstoff, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
L-Asparaginsäure, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Propylenglycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
Harnstoff, ACS reagent, 99.0-100.5%
Sigma-Aldrich
α-Linoleic acid, ≥98%
Supelco
Harnstoff, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Propylenglykol, meets USP testing specifications
Sigma-Aldrich
Harnstoff -Lösung, BioUltra, ~8 M in H2O
Supelco
Propylenglykol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1,2-Propandiol, ACS reagent, ≥99.5%
Sigma-Aldrich
Linolsäure, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Harnstoff, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Harnstoff, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Creatin, anhydrous
Sigma-Aldrich
Harnstoff, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
L-Asparaginsäure, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Tyramin, ≥98.0%
Sigma-Aldrich
Harnstoff, suitable for electrophoresis
Sigma-Aldrich
1,2-Propandiol, ReagentPlus®, 99%
Sigma-Aldrich
2,3-Butanediol, 98%
Sigma-Aldrich
Harnstoff, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
1,2-Propandiol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
L-Asparaginsäure, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
Linolsäure, technical, 58-74% (GC)
Supelco
Linolsäure, analytical standard
Sigma-Aldrich
Harnstoff, meets USP testing specifications
Sigma-Aldrich
L-Asparaginsäure, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Harnstoff, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
SAFC
L-Asparaginsäure
Sigma-Aldrich
Harnstoff -Lösung, 40 % (w/v) in H2O