- Inhibitor of apoptosis protein cIAP2 is essential for lipopolysaccharide-induced macrophage survival.
Inhibitor of apoptosis protein cIAP2 is essential for lipopolysaccharide-induced macrophage survival.
Molecular and cellular biology (2005-12-31)
Damiano Conte, Martin Holcik, Charles A Lefebvre, Eric Lacasse, David J Picketts, Kathryn E Wright, Robert G Korneluk
PMID16382159
ZUSAMMENFASSUNG
The cellular inhibitor of apoptosis 2 (cIAP2/HIAP1) is a potent inhibitor of apoptotic death. In contrast to the other members of the IAP family, cIAP2 is transcriptionally inducible by nuclear factor-kappaB in response to multiple triggers. We demonstrate here that cIAP2-/- mice exhibit profound resistance to lipopolysaccharide (LPS)-induced sepsis, specifically because of an attenuated inflammatory response. We show that LPS potently upregulates cIAP2 in macrophages and that cIAP2-/- macrophages are highly susceptible to apoptosis in a LPS-induced proinflammatory environment. Hence, cIAP2 is critical in the maintenance of a normal innate immune inflammatory response.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O127:B8, BioXtra, suitable for cell culture, γ-irradiated