Merck
Lack of protease activated receptor (PAR) expression in purified human basophils.
Inflammation research : official journal of the European Histamine Research Society ... [et al.] (2005-06-02)
F H Falcone, S Morroll, B F Gibbs
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
GenElute Säugetier-Gesamt-RNA-Miniprep-Kit, sufficient for 70 purifications
Sigma-Aldrich
GenElute Säugetier-Gesamt-RNA-Miniprep-Kit, sufficient for 350 purifications
Sigma-Aldrich
GenElute Säugetier-Gesamt-RNA-Miniprep-Kit, sufficient for 10 purifications