Merck
Interactions between new quinolone antibacterials and diagnostic drug containing manganese.
European journal of drug metabolism and pharmacokinetics (2013-02-28)
Moeko Hosono, Haruko Yokoyama, Risa Takayanagi, Yasuhiko Yamada
ZUSAMMENFASSUNG
A diagnostic drug containing manganese chloride tetrahydrate as a major ingredient is available since 2006. It is used in magnetic resonance imaging as a negative contrast medium for magnetic resonance cholangiopancreatography of the gastrointestinal tract. However, there is no report regarding interaction between manganese and new quinolone antibacterials. We investigated the interactions between new quinolone antibacterials and a diagnostic drug containing manganese in vitro. We evaluated the rate of formation of chelate complex by reacting new quinolone antibacterials (levofloxacin, ofloxacin, ciprofloxacin) with a diagnostic drug containing manganese. The EC50 values of the formation of chelate complex for levofloxacin, ofloxacin, and ciprofloxacin were 5.14 ± 0.14, 5.29 ± 0.14, and 0.96 ± 0.04 mM, respectively. The rates of formation of chelate complex by levofloxacin, ofloxacin, and ciprofloxacin in a reaction with the diagnostic drug were 17.0, 18.9, and 55.5 % in clinical condition, respectively. Our results suggest that a complex of each antibacterial and manganese was formed, with ciprofloxacin causing the strongest interaction. In addition, our findings indicate that the degree of interaction may be an important problem in clinical settings with concomitant administration of a new quinolone antibacterial and diagnostic drug containing manganese.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
Levofloxacin, 98.0-102.0% anhydrous basis (HPLC)
Sigma-Aldrich
Ofloxacin, fluoroquinolone antibiotic
Supelco
Ciprofloxacin HCl, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ciprofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Levofloxacin, analytical standard
Supelco
Ofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Ciprofloxacin-Hydrochlorid, European Pharmacopoeia (EP) Reference Standard
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Ciprofloxacin, European Pharmacopoeia (EP) Reference Standard
Supelco
Ofloxacin, VETRANAL®, analytical standard
Ciprofloxacinhydrochlorid für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Ofloxacin, European Pharmacopoeia (EP) Reference Standard