Merck
Ethylenediaminetetraacetic acid-dependent pseudomacrocytosis.
Journal of clinical pathology (2013-06-01)
Jose Manuel Vagace, Miguel Ángel Rodriguez, María Dolores de la Maya, Guillermo Gervasini
ZUSAMMENFASSUNG
We investigated the case of a 14-year-old girl with an ethylenediaminetetraacetic acid (EDTA)-dependent haemagglutination detected by macrocytosis, which was only evident by an abnormal red blood cell (RBC) population in the histogram. Investigations included haemograms with different anticoagulants and experimental conditions. Immunohaematological studies were performed using a gel-based technology. At admission, the patient had a low RBC count and an increased mean corpuscular volume with normal haemoglobin. A double population appeared in the RBC histogram. However, the peripheral blood smear was normal and macrocytosis was absent when heparin or citrate was used instead of EDTA. Later studies revealed that the patient's serum was able to induce macrocytosis of control RBC only in the presence of EDTA. An EDTA-dependent panagglutinin was then indentified that produced mixed field agglutination. These findings provide evidence of a haemagglutination induced by EDTA as a source of pseudomacrocytosis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, suitable for electrophoresis, Molecular Biology, 99.0-101.0% (titration)
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, ACS reagent, 99.0-101.0%
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, Molecular Biology, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Supelco
Dinatriumedetat Dihydrat, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, Sigma Grade, suitable for plant cell culture, 98.5-101.5%
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
Sigma-Aldrich
Ethylendiamintetraessigsäure Calcium-Dinatriumsalz Hydrat
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, 2%, solution
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, BioUltra, 98.5-101.5%
Sigma-Aldrich
Ethylendiamintetraessigsäure, ≥98.0% (KT)
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, ≥97.0% (KT)