Merck
Xanthine oxidase inhibitory lanostanoids from Ganoderma tsugae.
Fitoterapia (2013-06-19)
Kai-Wei Lin, Yen-Ting Chen, Shyh-Chyun Yang, Bai-Luh Wei, Chi-Feng Hung, Chun-Nan Lin
ZUSAMMENFASSUNG
Two new lanostanoids, 3α-acetoxy-22-oxo-5α-lanosta-8,24-dien-21-oic acid, named tsugaric acid D (1) and 16α-hydroxy-3-oxo-5α-lanosta-6,8,24(24(1))-trien-21-oic acid, named tsugaric acid E (2) were isolated from the fruit bodies of Ganoderma tsugae. The structures 1 and 2 were determined by spectroscopic methods. Compound 1 and known compounds 3 and 6 exhibited significant inhibitory effects on xanthine oxidase (XO) activity with an IC50 values of 90.2±24.2, 116.1±3.0, and 181.9±5.8 μM, respectively. Known compound 5 was able to protect human keratinocytes against damage induced by UVB light, which showed 5 could protect keratinocytes from photodamage. The 1 and 5 μM 1 combined with 5 μM cisplatin, respectively, enhanced the cytotoxicity induced by cisplatin. It suggested that 1 and 5 μM 1 combined with low dose of cisplatin may enhance the therapeutic efficacy of cisplatin and reduce side effect and cisplatin resistant.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Xanthinoxidase aus Kuhmilch, Grade I, ammonium sulfate suspension, ≥0.4 units/mg protein
Sigma-Aldrich
Xanthinoxidase aus Kuhmilch, lyophilized powder, 0.4-1.0 units/mg protein
Sigma-Aldrich
Xanthinoxidase aus microbiell, lyophilized powder, ≥7 units/mg solid
Sigma-Aldrich
Xanthinoxidase aus Kuhmilch, Grade IV, ammonium sulfate suspension, ≥0.1 units/mg protein
Sigma-Aldrich
Lanosterin, ≥93%, powder
Sigma-Aldrich
Xanthinoxidase aus Kuhmilch, Grade III, ammonium sulfate suspension, ≥0.8 units/mg protein