Merck
  • Preparative isolation and purification of five steroid saponins from Dioscorea zingiberensis C.H.Wright by counter-current chromatography coupled with evaporative light scattering detector.
Preparative isolation and purification of five steroid saponins from Dioscorea zingiberensis C.H.Wright by counter-current chromatography coupled with evaporative light scattering detector.
Journal of pharmaceutical and biomedical analysis (2013-07-09)
Xinxin Zhang, Yoichiro Ito, Jinru Liang, Qi Su, Yongmin Zhang, Jianli Liu, Wenji Sun
ZUSAMMENFASSUNG
A counter-current chromatography (CCC) method was successfully applied to separate and purify steroid saponins from the traditional Chinese medicine Dioscorea zingiberensis C.H.Wright for the first time. Ethyl acetate-n-butanol-methanol-water (4:1:2:4, v/v) was used as the two-phase solvent system, and evaporative light scattering detector (ELSD) was used as the detector in this method. The method separated in a single run the following five steroid saponins: 26-O-β-d-glucopyranosyl-(25R)-furost-5-en-3β, 22ζ, 26-triol-3-O-[β-d-glucopyranosyl-(1→3)-β-d-glucopyranol-(1→4)-α-l-rhamnopyranosyl-(1→2)]-β-d-glucopyranoside (Compound A); 26-O-β-d-glucopyranosyl-(25R)-furost-5-en-3β, 22ζ, 26-triol-3-O-[β-d-glucopyranosyl(1→3)-α-l-rhamnopyranosyl(1→2)]-β-d-glucopyranoside (Compound B); 26-O-β-d-glucopyranosyl-(25R)-furost-5-en-3β, 22ζ, 26-triol-3-O-[α-l-rhamnopyranosyl(1→4)]-β-d-glucopyranoside (Compound C); 26-O-β-d-glucopyranosyl-(25R)-furost-5, 20(22)-diene-3β, 26-diol-3-O-{α-l-rhamnopyranosyl-(1→4)-[β-d-glucopyranosyl-(1→3)-β-d-glucopyranosyl-(1→2)]}-β-d-glucopyranoside (Compound D); and 26-O-β-d-glucopyranosyl-(25R)-furost-5, 20(22)-diene-3β, 26-diol-3-O-[β-d-glucopyranosyl-(1→4)-α-l-rhamnopyranosyl(1→2)]-β-d-glucopyranoside (Compound E). Their structural identification of the five steroid saponins was performed by means of ESI-MS, and (13)C NMR.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethylacetat, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethylacetat, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
1-Butanol, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethylacetat, anhydrous, 99.8%
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Supelco
Ethylacetat, analytical standard
Sigma-Aldrich
1-Butanol, Molecular Biology, ≥99%
Sigma-Aldrich
Ethylacetat, ≥99%, FCC, FG
Sigma-Aldrich
Ethylacetat, ACS reagent, ≥99.5%
Sigma-Aldrich
Butylalkohol, natural, ≥99.5%, FCC, FG
Supelco
1-Butanol, analytical standard
Sigma-Aldrich
Ethylacetat, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethylacetat, biotech. grade, ≥99.8%
Sigma-Aldrich
Ethylacetat, ReagentPlus®, ≥99.8%
Sigma-Aldrich
1-Butanol, BioRenewable, ACS reagent, ≥99.4%