Merck
  • Comparison of the behavior of fibroblast and bone marrow-derived mesenchymal stem cell on nitrogen plasma-treated gelatin films.
Comparison of the behavior of fibroblast and bone marrow-derived mesenchymal stem cell on nitrogen plasma-treated gelatin films.
Materials science & engineering. C, Materials for biological applications (2013-08-06)
I Prasertsung, S Kanokpanont, R Mongkolnavin, C S Wong, J Panpranot, S Damrongsakkul
ZUSAMMENFASSUNG
The attachment and growth behavior of mouse fibroblast (L929) and rat bone marrow-derived mesenchymal stem cell (MSC) on nitrogen plasma-treated and untreated gelatin films was investigated and compared. The gelatin films were prepared by solution casting (0.05% w/v) and crosslinked using dehydrothermal treatment. The crosslinked gelatin films were treated with nitrogen alternating current (AC) 50 Hz plasma systems at various treatment time. The results on the attachment and growth of two cells; L929 and MSC, on plasma-treated gelatin film showed that the number of attached and proliferated cells on plasma-treated gelatin films was significantly increased compared to untreated samples. However, no significant difference between the number of attached L929 and MSC on plasma-treated gelatin was observed. The shorter population doubling time and higher growth rate of cells cultured on plasma-treated film indicated the greater growth of cells, compared to ones on untreated films. The greatest enhancement of cell attachment and growth were noticed when the film was treated with nitrogen plasma for 9 to 15s. This suggested that the greater attachment and growth of both cells on gelatin films resulted from the change of surface properties, i.e. hydrophilicity, surface energy, and chemistry. The suitable water contact angle and oxygen/nitrogen ratio (O/N) of gelatin film for best L929 and MSC attachment were observed at 27-32° and 1.4, respectively. These conditions also provided the best proliferation of cells on plasma-treated gelatin films.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Gelatine -Lösung, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture
Sigma-Aldrich
Gelatine aus Schweinehaut, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture
Sigma-Aldrich
Gelatine aus Schweinehaut, gel strength 300, Type A
Sigma-Aldrich
Gelatine aus Haut von Kaltwasserfischen, solid
Sigma-Aldrich
Gelatine aus Rinderhaut, Type B, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Gelatine aus Rinderhaut, gel strength ~225 g Bloom, Type B
Sigma-Aldrich
Gelatine aus Schweinehaut, gel strength ~175 g Bloom, Type A
Sigma-Aldrich
Gelatine aus Rinderhaut, Type B
Sigma-Aldrich
Gelatine aus Schweinehaut, gel strength 80-120 g Bloom, Type A
Sigma-Aldrich
Gelatine, tested according to Ph. Eur.
Sigma-Aldrich
Prionex® Highly purified Type A, aqueous solution
Millipore
Gelatine aus Schweinehaut, medium gel strength, suitable for microbiology
Sigma-Aldrich
Gelatine aus Schweinehaut, Type A, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture
Millipore
Gelatine aus Schweinehaut, suitable for microbiology, high gel strength
Millipore
Gelatine aus Schweinehaut, suitable for microbiology, ultrahigh gel strength
Millipore
Gelatine aus Schweinehaut, suitable for microbiology, low gel strength