Merck
Red scrotum syndrome: idiopathic neurovascular phenomenon or steroid addiction?
Sexual health (2013-09-14)
Tarun Narang, Muthu Sendhil Kumaran, Sunil Dogra, Uma Nahar Saikia, Bhushan Kumar
ZUSAMMENFASSUNG
Red scrotum syndrome (RSS) is not infrequent but is often misdiagnosed or underdiagnosed, and seldom reported. The exact etiopathogeneis is still unknown but it almost always follows the prolonged application of topical corticosteroids and is characterised by persistent erythema of the scrotum, associated with severe itching, hyperalgesia and a burning sensation. To evaluate the clinicoepidemiological profile and assess the efficacy of various treatment modalities in addition to corticosteroid abstinence in the treatment of RSS. Twelve patients with RSS, who presented to us during 2010 and 2011, were identified, and various aspects of their illness and treatment were studied. Patch testing was performed in all patients. A skin biopsy was done in seven patients. The average age of the patients was 45.83 years (26-62 years). The average duration of illness or the duration of topical steroid use was 27.41 months (6-56 months). Psychiatric comorbidities were seen in 9 (75%) out of 12 patients. Histopathology revealed features resembling erythematotelengiectatic rosacea in four of the biopsied patients. Patch test results were negative. All patients reported improvement of their symptoms within 4 weeks of starting doxycycline with amitriptyline or pregabalin; the treatment had to be continued for 3-4 months. RSS appears to be a manifestation of corticosteroid misuse rather than a primary disease. We suggest that RSS is a rosacea-like dermatosis or steroid-induced rebound vasodilation based on clinical and histopathological features. Our patients responded to cessation of steroids and doxycycline in combination with amitryptaline or pregabalin.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Doxycyclin -hyclat
Sigma-Aldrich
Amitriptylin -hydrochlorid, ≥98% (TLC), powder
Supelco
Doxycyclin -hyclat, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Doxycycline monohydrate
Supelco
Amitriptyline hydrochloride solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Doxycyclin -hyclat, VETRANAL®, analytical standard