Merck
  • Endogenous glucocorticoid receptor signaling drives rhythmic changes in human T-cell subset numbers and the expression of the chemokine receptor CXCR4.
Endogenous glucocorticoid receptor signaling drives rhythmic changes in human T-cell subset numbers and the expression of the chemokine receptor CXCR4.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2013-09-21)
Luciana Besedovsky, Jan Born, Tanja Lange
ZUSAMMENFASSUNG
In humans, numbers of circulating naive T cells strongly decline in the morning, which was suggested to be mediated by cortisol, inducing a CXCR4 up-regulation with a subsequent extravasation of the cells. As a systematic evaluation of this assumption is lacking, we investigated in two human placebo-controlled studies the effects of the glucocorticoid receptor (GR) antagonist mifepristone (200 mg orally at 23:00) and of suppressing endogenous cortisol with metyrapone (1 g orally at 04:00) on temporal changes in CXCR4 expression and numbers of different T-cell subsets using flow cytometry. Mifepristone attenuated, and metyrapone completely blocked, the morning increase in CXCR4 expression on naive T cells. In parallel, both substances also hindered the decline in naive T-cell numbers with this effect, however, being less apparent after mifepristone. We identified, and confirmed in additional in vitro studies, a partial agonistic GR effect of mifepristone at night (i.e., between 02:00 and 03:30) that could explain the lower antagonistic efficacy of the substance on CXCR4 expression and naive T-cell counts. CXCR4 expression emerged to be a most sensitive marker of GR signaling. Our studies jointly show that endogenous cortisol, specifically via GR activation, causes the morning increase in CXCR4 expression and the subsequent extravasation of naive T cells, thus revealing an important immunological function of the morning cortisol rise.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Mifepriston, ≥98%
Sigma-Aldrich
2-Methyl-1,2-di-3-Pyridyl-1-Propanon, 96%
Sigma-Aldrich
Metyrapon, ≥98% (HPLC), solid