Merck
  • The effectiveness and risk comparison of EDTA with EGTA in enhancing Cd phytoextraction by Mirabilis jalapa L.
The effectiveness and risk comparison of EDTA with EGTA in enhancing Cd phytoextraction by Mirabilis jalapa L.
Environmental monitoring and assessment (2013-09-27)
Song Wang, Jianv Liu
ZUSAMMENFASSUNG
In the previous study, Mirabilis jalapa L. had revealed the basic Cd hyperaccumulator characteristics, but the accumulation ability was not as strong as that of other known Cd hyperaccumulators. In order to improve the accumulation ability of this ornamental plant, the chelants were used to activate the Cd in soil. As a substitute, ethylene glycol bis(2-aminoethyl) tetraacetic acid (EGTA) was selected to testify whether it has better effectiveness and can bring lesser metal leaching risk than EDTA. The data showed that the growth of M. jalapa was inhibited, while the Cd concentration of the plant was significantly increased under the treatments containing EDTA or EGTA. The Cd translocation ability under the EGTA treatments was higher than that under the EDTA treatments. The available Cd resulted from the application of chelant EGTA to the contaminated soils can be limited to the top 5 cm, while the application of chelant EDTA to the contaminated soils can be limited to the top 10 cm. In a word, EGTA showed better effectiveness than EDTA in enhancing Cd phytoextraction of M. jalapa. As an ornamental plant, M. jalapa has the potential to be used for phytoextraction of Cd-contaminated soils and it can beautify the environment at the same time.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, suitable for electrophoresis, Molecular Biology, 99.0-101.0% (titration)
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, ACS reagent, 99.0-101.0%
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenglykol-bis(2-aminoethylether)-N,N,N′,N′-Tetraessigsäure, Molecular Biology, ≥97.0%
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, Molecular Biology, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Supelco
Dinatriumedetat Dihydrat, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylenglykol-bis(2-aminoethylether)-N,N,N′,N′-Tetraessigsäure, ≥97.0%
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, Sigma Grade, suitable for plant cell culture, 98.5-101.5%
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
Sigma-Aldrich
Ethylendiamintetraessigsäure Calcium-Dinatriumsalz Hydrat
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ethylenglykol-bis(2-aminoethylether)-N,N,N′,N′-Tetraessigsäure, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, 2%, solution
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, BioUltra, 98.5-101.5%
Sigma-Aldrich
Ethylendiamintetraessigsäure, ≥98.0% (KT)
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Ethylenglykol-bis(2-aminoethylether)-N,N,N′,N′-Tetraessigsäure, BioXtra, ≥97 .0%
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, ≥97.0% (KT)
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis