Merck
A Dictyostelium cellobiohydrolase orthologue that affects developmental timing.
Development genes and evolution (2013-11-19)
Mizuho Kunii, Mami Yasuno, Yuki Shindo, Takefumi Kawata
ZUSAMMENFASSUNG
Dictyostelium discoideum is a facultative multicellular amoebozoan with cellulose in the stalk and spore coat of its fruiting body as well as in the extracellular matrix of the migrating slug. The organism also harbors a number of cellulase genes. One of them, cbhA, was identified as a candidate cellobiohydrolase gene based on the strong homology of its predicted protein product to fungal cellobiohydrolase I (CBHI). Expression of the cbhA was developmentally regulated, with strong expression in the spores of the mature fruiting body. However, a weak but detectable level of expression was observed in the extracellular matrix at the mound - tipped finger stages, in prestalk O cells, and in the slime sheath of the migrating slug - late culminant stages. A null mutant of the cbhA showed almost normal morphology. However, the developmental timing of the mutant was delayed by 2-4 h. When a c-Myc epitope-tagged CbhA was expressed, it was secreted into the culture medium and was able to bind crystalline cellulose. The CbhA-myc protein was glycosylated, as demonstrated by its ability to bind succinyl concanavalin A-agarose. Moreover, conditioned medium from the cbhA-myc (oe) strain displayed 4-methylumbelliferyl β-D-cellobioside (4-MUC) digesting activity in Zymograms in which conditioned medium was examined via native-polyacrylamide gel electrophoresis or spotted on an agar plate containing 4-MUC, one of the substrates of cellobiohydrolase. Taken together, these findings indicate that Dictyostelium CbhA is an orthologue of CBH I that is required for a normal rate of development.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cellulosepulver, microcrystalline, powder, 20 μm
Sigma-Aldrich
Cellulosepulver, microcrystalline, powder
Sigma-Aldrich
Cellulosepulver, fibers, (medium)
Sigma-Aldrich
Sigmacell Cellulose, Type 20, 20 μm
Supelco
Avicel® PH-101, ~50 μm particle size
Sigma-Aldrich
α-Cellulose, powder
Sigma-Aldrich
Cellulosepulver, colloidal, microcrystalline
Sigma-Aldrich
Sigmacell Cellulose, Type 50, 50 μm
Sigma-Aldrich
Sigmacell Cellulose, Type 101, Highly purified, fibers
Sigma-Aldrich
Avicel® PH-101, tested according to Ph. Eur.
Sigma-Aldrich
α-Cellulose, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Cellobiohydrolase I from Hypocrea jecorina, 100 U/mg, recombinant, expressed in corn
Supelco
Cellulosepulver, powder, suitable for column chromatography
Supelco
Cellulosepulver, DS-0, powder, suitable for thin layer chromatography (TLC)
Supelco
Cellulosepulver, acid washed, powder, suitable for column chromatography
Supelco
Cellulosepulver, DFS-0, microcrystalline, suitable for thin layer chromatography (TLC)
Supelco
Cellulosepulver, acid washed, from spruce, suitable for column chromatography